Skip to Content
Merck
All Photos(8)

Documents

HPA018864

Sigma-Aldrich

Anti-FOXK1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Forkhead box protein K1, Anti-MNF, Anti-Myocyte nuclear factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

GVPGHTVTILQPATPVTLGQHHLPVRAVTQNGKHAVPTNSLAGNAYALTSPLQLLATQASSSAPVVVTRVCEVGPKEPAAAVAATATTTPATATTASASASSTGEPEVKRSRVEEPSGAVTTPAG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOXK1(221937)

General description

The gene FOXK1 (forkhead box protein K1) is mapped to human chromosome 7p22.1. It belongs to forkhead-box (FOX) family transcription factors. FOXK1 is present in immature tissues of brain, eye, heart, lung and thymus. However, in adults FOXK1 is particularly present in malignant tissues (brain, colon and lymph node).

Immunogen

Forkhead box protein K1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Forkhead box protein K1 (FOXK1) is involved in carcinogenesis and embryogenesis. FOXK1 is up-regulated in colorectal cancers. It binds dishevelled (DVL) and translocates it to the nucleus, thereby activating Wnt/β-catenin signaling. FOXK1 regulates many genes involved in cell cycle, including DHFR (dihydrofolate reductase), TYMS (thymidylate synthase), GSDMD (gasdermin-D), and TFDP1 (transcription factor Dp-1). Absence of FOXK1 decreases cell proliferation rates.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73353

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenqi Wang et al.
Developmental cell, 32(6), 707-718 (2015-03-26)
Dishevelled (DVL) proteins serve as crucial regulators that transduce canonical Wnt signals to the GSK3β-destruction complex, resulting in the stabilization of β-catenin. Emerging evidence underscores the nuclear functions of DVLs, which are critical for Wnt/β-catenin signaling. However, the mechanism underlying
Masuko Katoh et al.
International journal of molecular medicine, 14(1), 127-132 (2004-06-18)
Forkhead-box (FOX) family transcription factors are implicated in carcinogenesis and embryogenesis. Here, we identified and characterized the human FOXK1 gene by using bioinformatics. Complete coding sequence of human FOXK1 cDNA was determined by assembling CB959941 EST, AW206906 EST, and 5'-truncated
Gavin D Grant et al.
Molecular biology of the cell, 23(16), 3079-3093 (2012-06-29)
We developed a system to monitor periodic luciferase activity from cell cycle-regulated promoters in synchronous cells. Reporters were driven by a minimal human E2F1 promoter with peak expression in G1/S or a basal promoter with six Forkhead DNA-binding sites with
Jeffrey T J Huang et al.
International journal of oncology, 25(3), 751-757 (2004-08-04)
We describe here the identification and characterization of a human forkhead box gene, FOXK1, that encodes predicted proteins most homologous to the mouse myocyte nuclear factor (MNF)/forkhead box K1 (FoxK1). Human FOXK1 is located at the chromosomal position 7p22. Two
Fucun Gao et al.
OncoTargets and therapy, 13, 623-634 (2020-02-06)
Forkhead box K1 (FOXK1) is members of the FOX transcription factor family. Previous work has found out that FOXK1 promotes cell proliferation, migration and invasion in several cancers, such as gastric cancer, glioma cancer and lung cancer; however, the exact

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service