Skip to Content
Merck
All Photos(6)

Key Documents

HPA011175

Sigma-Aldrich

Anti-MANF antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ARMET, Anti-Arginine-rich protein, Anti-MANF, Anti-Mesencephalic astrocyte-derived neurotrophic factor, Anti-Protein ARMET precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

CEVCISYLGRFYQDLKDRDVTFSPATIENELIKFCREAR

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARMET(7873)

General description

MANF (mesencephalic astrocyte-derived neurotrophic factor) forms a family of neurotrophic factors (NTFs) along with cerebral dopamine neurotrophic factor (CDNF). It is characterized by eight conserved cysteine residues, a saposin-like domain in the N-terminal and a putative unfolded carboxy-terminal domain. This protein has a 21 amino acids long signal peptide, which is cleaved off to yield a mature protein of 158 amino acids. MANF is a secreted protein, but some of it remains in endoplasmic reticulum (ER). It was initially isolated from cultured embryonic dopaminergic neurons, and in brain, is predominantly expressed in cerebral cortex, hippocampus and cerebellar Purkinje cells. This gene is localized to human chromosome 3, spans 4.3kb, and contains 4 exons. The encoded protein has a molecular weight of 20kDa.

Immunogen

Protein ARMET precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MANF (mesencephalic astrocyte-derived neurotrophic factor) plays a protective role towards midbrain dopaminergic neurons. It is an endoplasmic reticulum (ER) stress related protein. Its levels are increased post brain injury, ischemia and epileptic insults. It might confer neuro-protection against brain ischemia, by preventing necrosis or apoptosis of cerebral cortex cells. It is over-expressed during unfolded protein response (UPR), and prevents ER stress-induced apoptosis, as well as cell proliferation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72053

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Penka Petrova et al.
Journal of molecular neuroscience : MN, 20(2), 173-188 (2003-06-10)
We describe the discovery of a novel, 20 kDa, secreted human protein named mesencephalic astrocyte-derived neurotrophic factor, or MANF. The homologous, native molecule was initially derived from a rat mesencephalic type-1 astrocyte cell line and recombinant MANF subcloned from a
Mikko Airavaara et al.
Experimental neurology, 225(1), 104-113 (2010-08-06)
Mesencephalic astrocyte-derived neurotrophic factor (MANF) is a secreted protein which reduces endoplasmic reticulum (ER) stress and has neurotrophic effects on dopaminergic neurons. Intracortical delivery of recombinant MANF protein protects tissue from ischemic brain injury in vivo. In this study, we
Päivi Lindholm et al.
Developmental neurobiology, 70(5), 360-371 (2010-02-27)
Current therapeutic interventions for neurodegenerative diseases alleviate only disease symptoms, while treatments that could stop or reverse actual degenerative processes are not available. Parkinson's disease (PD) is a movement disorder with characteristic degeneration of dopaminergic neurons in the midbrain. Few
Andria Apostolou et al.
Experimental cell research, 314(13), 2454-2467 (2008-06-20)
The accumulation of misfolded proteins in the endoplasmic reticulum (ER) causes ER stress that initiates the unfolded protein response (UPR). UPR activates both adaptive and apoptotic pathways, which contribute differently to disease pathogenesis. To further understand the functional mechanisms of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service