Skip to Content
Merck
All Photos(5)

Documents

HPA011166

Sigma-Aldrich

Anti-ATP2B1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PMCA1, Anti-Plasma membrane calcium ATPase isoform 1, Anti-Plasma membrane calcium pump isoform 1, Anti-Plasma membrane calcium-transporting ATPase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

DYQDVRNEIPEEALYKVYTFNSVRKSMSTVLKNSDGSYRIFSKGASEIILKKCFKILSANGEAKVFRPRDRDDIVKTVIEPMASEGLRTICLAFRDFPAGEPEPEWDNENDIVTGLTCIAVVGI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATP2B1(490)

Immunogen

Plasma membrane calcium-transporting ATPase 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ATP2B1 (ATPase, Ca++ transporting, plasma membrane 1) is responsible for the efflux of Ca2+ ions from the cells, against substantial concentration gradients. Therefore, it is responsible for maintaining Ca2+ homeostasis in the cell. It pushes Ca2+ ions out of the cells, against the gradient by using the energy derived from the hydrolysis of ATP. It also plays a role in Ca2+ signaling in the cell. Ultraviolet B irradiation decreases expression of this protein, which might be responsible for aberrant intracellular Ca2+ levels, and hence apoptosis of human lens epithelial cells. Studies show that ATP2B1 plays a role in regulating blood pressure (BP) by modulating vasoconstriction and Ca2+ levels in vascular smooth muscle cells. Genome wide association studies have linked this gene to hypertension.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71543

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiuxin Wu et al.
Molecular and cellular biochemistry, 382(1-2), 263-272 (2013-07-03)
Ultraviolet B (UVB) could lead to the apoptosis of human lens epithelial cell and be hypothesized to be one of the important factors of cataractogenesis. In the human lens, Ca(2+)-ATPase is a major determinant of calcium homeostasis. Plasma membrane calcium
Yan Wang et al.
Molecular biology reports, 40(2), 1867-1873 (2012-10-20)
Genome-wide association studies have identified the ATP2B1 gene associated with blood pressure (BP), the evidence from large scale Chinese population was still rare. We performed the current replication study to test the association of the ATP2B1 gene and hypertension and
Nadine Schmidt et al.
Neuron, 96(4), 827-838 (2017-10-24)
Plasma membrane Ca2+-ATPases (PMCAs), a family of P-type ATPases, extrude Ca2+ ions from the cytosol to the extracellular space and are considered to be key regulators of Ca2+ signaling. Here we show by functional proteomics that native PMCAs are heteromeric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service