Skip to Content
Merck
All Photos(3)

Documents

HPA010645

Sigma-Aldrich

Anti-SAMSN1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Hematopoietic adaptor containing SH3 and SAM domains 1, Anti-SAM domain, SH3 domain and nuclear localization signals protein 1, Anti-SAM domain-containing protein SAMSN-1, Anti-SH3-SAM adaptor protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

TEAHEGDPTNGSGEQSKTSNNGGGLGKKMRAISWTMKKKVGKKYIKALSEEKDEEDGENAHPYRNSDPVIGTHTEKVSLKASDSMDSLYSGQSSSSGITSCSDGTSNRDSFRLDDDGPYSGPFC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SAMSN1(64092)

General description

SAMSN1 (SAM domain, SH3 domain and nuclear localization signals 1) gene encodes a member of the SLY family of cytoplasmic adaptor proteins. It is abundantly expressed in the hematopoietic compartment. The protein is also expressed in heart, brain, placenta, and lung to a lesser extent. The members of this family are characterized by the presence of protein-protein interaction domains and are homologous. They contain a sterile α motif (SAM) domain that is capable of self-association and can also associate with each other. The chimeric fusion of this domain with the βPDGF receptor, AML1, c-Abl, and JAK2 results in the oncogenic transformation of the SAM domain. The SH3 domains mediate protein-protein interactions in several signal transduction pathways. The gene is localized to human chromosome 21 (21q11.2).

Immunogen

SAM domain-containing protein SAMSN-1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SAMSN1 (SAM domain, SH3 domain and nuclear localization signals 1) protein may be involved in the formation of blood vessel during normal brain development. It may function as an adaptor or scaffolding protein. It has been found to be overexpressed in glioma and is associated with a poor survival in glioblastoma multiforme. In mice, this protein may function in moderating adaptive immune responses. SAMSN1 may be involved in B cell activation and differentiation. It has been suggested to be a tumor suppressor in lung cancer. It may serve as a biomarker of the phenotype of human hepatocellular carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71504

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoshi Sueoka et al.
Annals of surgical oncology, 22 Suppl 3, S1453-S1460 (2015-03-26)
Identification of molecular markers for sensitive detection of hepatocellular carcinoma (HCC) is required to achieve efficacious personalized therapy. We focused here on SAM domain, SH3 domain, and nuclear localization signals 1 (SAMSN1) and investigated expression and methylation status of SAMSN1
Yong Yan et al.
PloS one, 8(11), e81905-e81905 (2013-11-28)
To study the expression pattern and prognostic significance of SAMSN1 in glioma. Affymetrix and Arrystar gene microarray data in the setting of glioma was analyzed to preliminarily study the expression pattern of SAMSN1 in glioma tissues, and Hieratical clustering of
Natasha L Friend et al.
FASEB bioAdvances, 2(9), 554-572 (2020-09-15)
The protein SAMSN1 was recently identified as a putative tumor suppressor in multiple myeloma, with re-expression of Samsn1 in the 5TGM1/KaLwRij murine model of myeloma leading to a near complete abrogation of intramedullary tumor growth. Here, we sought to clarify
Jacqueline E Noll et al.
Neoplasia (New York, N.Y.), 16(7), 572-585 (2014-08-15)
Multiple myeloma (MM), a hematological malignancy characterized by the clonal growth of malignant plasma cells (PCs) in the bone marrow, is preceded by the benign asymptomatic condition, monoclonal gammopathy of undetermined significance (MGUS). Several genetic abnormalities have been identified as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service