Skip to Content
Merck
All Photos(3)

Documents

HPA005697

Sigma-Aldrich

Anti-LDOC1L antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-leucine zipper, down-regulated in cancer 1-like antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SVMAELTLLRTRARIPGALQITPPISSITSNGTRPMTTPPTSLPEPFSGDPGRLAGFLMQMDRFMIFQASRFPGEAERVAFLVSRLTGEAEKWAIPHMQPDSPLRNNYQGF

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LDOC1L(84247)

Related Categories

General description

The gene encoding leucine zipper protein down-regulated in cancer cells-like 1 (LDOC1L) is present on chromosome Xq27. LDOC1L is a 17kDa protein that possesses a leucine zipper-like motif at the N-terminal region and a short proline-rich region containing SH3-binding consensus sequence.

Immunogen

leucine zipper, down-regulated in cancer 1-like recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Leucine zipper protein down-regulated in cancer cells-like 1 (LDOC1L) is a tumor suppressor and novel regulator of the transcriptional response mediated by the nuclear factor kappa B (NF-κβ). It is also involved in apoptosis, wherein it plays an important role in the externalization of the cell membrane phosphatidylserine and reduces cell viability. This apoptosis is accompanied by an increase in the p53 protein level and not in p53 transcription showing that LDOC1 inhibits the degradation of p53.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73621

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kiyohito Mizutani et al.
Journal of biochemistry, 138(5), 639-646 (2005-11-08)
WAVE3 belongs to the Wiskott-Aldrich syndrome protein family that mediates actin reorganization through activation of the Arp2/3 complex. However, the physiological function of WAVE3 is poorly understood. We found that LDOC1-encoded by a gene that is down-regulated in tumor cell
Koichi Nagasaki et al.
International journal of cancer, 105(4), 454-458 (2003-04-25)
We have isolated a novel gene, LDOC1, which encodes for a leucine zipper protein that was downregulated in a series of human pancreatic cancer cell lines but was expressed in corresponding normal tissues. We report the initial characterization of LDOC1
Mikiya Inoue et al.
FEBS letters, 579(3), 604-608 (2005-01-27)
LDOC1 was isolated as a gene encoding a leucine-zipper protein whose expression was decreased in pancreatic and gastric cancer cell lines in 1999. Here, we found that overexpression of LDOC1 caused externalization of the cell membrane phosphatidylserine, which was characteristic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service