Skip to Content
Merck
All Photos(2)

Documents

HPA002835

Sigma-Aldrich

Anti-TROVE2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Ro60, Anti-SSA2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:1000-1:2500
western blot: 0.04-0.4 μg/mL

immunogen sequence

FLLAVDVSASMNQRVLGSILNASTVAAAMCMVVTRTEKDSYVVAFSDEMVPCPVTTDMTLQQVLMAMSQIPAGGTDCSLPMIWAQKTNTPADVFIVFTDNETFAGGVHPAIALREYRKKMDIPAKLIVCGMTSNGF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TROVE2(6738)

General description

TROVE2 is a 60kDa SS (single stranded)-A/Ro ribonucleoprotein consisting of an α-helical HEAT repeat region. This part forms a ring with a central hole, which offer RNA binding surface to a large collection of possible RNAs, including Y RNAs, variant 5S rRNAs, and misfolded U2 small nuclear RNA.

Immunogen

60 kDa SS-A/Ro ribonucleoprotein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

60kDa SS-A/Ro ribonucleoprotein is a protein encoded by the TROVE2 gene in humans. The gene is also known as RO60, SSA2 and RORNP. The gene binds noncoding Y RNA and only becomes accessible to autoantibodies during apoptosis. It may play an important role in trafficking the development of cardiac neonatal lupus (NL). It also imparts a critical role in the pathogenicity of maternal anti-Ro60 autoantibodies.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86207

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryota Naito et al.
JCI insight, 8(16) (2023-07-11)
The U1RNP complex, Ro/SSA, and La/SSB are major RNA-containing autoantigens. Immune complexes (ICs) composed of RNA-containing autoantigens and autoantibodies are suspected to be involved in the pathogenesis of some systemic autoimmune diseases. Therefore, RNase treatment, which degrades RNA in ICs
Joanne H Reed et al.
Journal of immunology (Baltimore, Md. : 1950), 191(1), 110-116 (2013-05-24)
Cardiac neonatal lupus (NL) is presumed to arise from maternal autoantibody targeting an intracellular ribonucleoprotein, Ro60, which binds noncoding Y RNA and only becomes accessible to autoantibodies during apoptosis. Despite the importance of Ro60 trafficking in the development of cardiac
Robert M Clancy et al.
Journal of immunology (Baltimore, Md. : 1950), 184(4), 2148-2155 (2010-01-22)
Activation of TLR by ssRNA after FcgammaR-mediated phagocytosis of immune complexes (IC) may be relevant in autoimmune-associated congenital heart block (CHB) where the obligate factor is a maternal anti-SSA/Ro Ab and the fetal factors, protein/RNA on an apoptotic cardiocyte and
CoLoC-seq probes the global topology of organelle transcriptomes.
Jeandard, et al.
Nucleic Acids Research, 51, e16-e16 (2023)
Bin Yang et al.
Proteomics, 15(15), 2602-2617 (2015-03-18)
Brain-enriched miR-128 is repressed in glioma cells, and could inhibit the proliferation of gliomas by targeting genes such as E2F3a and BMI1. To identify more targets of miR-128 in glioblastoma multiforme, the pulse stable isotope labeling with amino acids in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service