Skip to Content
Merck
All Photos(2)

Documents

HPA001432

Sigma-Aldrich

Anti-HSD17B10 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-3-Hydroxy-2-methylbutyryl-CoA dehydrogenase antibody produced in rabbit, Anti-3-Hydroxyacyl-CoA dehydrogenase type II antibody produced in rabbit, Anti-3-Hydroxyacyl-CoA dehydrogenase type-2 antibody produced in rabbit, Anti-Endoplasmic reticulum-associated amyloid β-peptide-binding protein antibody produced in rabbit, Anti-Type II HADH, Anti-Type II HADH antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

EAQAKKLGNNCVFAPADVTSEKDVQTALALAKGKFGRVDVAVNCAGIAVASKTYNLKKGQTHTLEDFQRVLDVNLMGTFNVIRLVAGEMGQNEPDQGGQRGVIINTASVAAFEGQVGQA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSD17B10(3028)

General description

HSD17B10 (hydroxysteroid (17-b) dehydrogenase 10) gene encodes a mitochondrial, multifunctional protein, 17β-hydroxysteroid dehydrogenase type 10. It is a homotetrameric protein belonging to the short-chain dehydrogenase/reductase superfamily.

Immunogen

3-Hydroxyacyl-CoA dehydrogenase type-2 recombinant protein epitope signature tag (PrEST)

Application

Anti-HSD17B10 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

HSD17B10 (hydroxysteroid (17-β) dehydrogenase 10) is involved in the metabolism of fatty acids and steroid. It catalyzes the 2-methyl-3-hydroxybutyryl-CoA dehydrogenation (MHBD) reaction in isoleucine metabolism. It is a subunit of mitochondrial ribonuclease P and functions in the maturation of mitochondrial tRNA. It catalyzes the oxidation of several fatty acids and steroids. Defects in this gene cause 17β-hydroxysteroid dehydrogenase type 10 (HSD10) deficiency, that is characterized by progressive psychomotor regression and alteration of mitochondria morphology. It has been reported that HSD17B10 shows clinical importance in Alzheimer disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74041

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Song-Yu Yang et al.
The Journal of steroid biochemistry and molecular biology, 143, 460-472 (2014-07-11)
17β-Hydroxysteroid dehydrogenase type 10 (17β-HSD10) is encoded by the HSD17B10 gene mapping at Xp11.2. This homotetrameric mitochondrial multifunctional enzyme catalyzes the oxidation of neuroactive steroids and the degradation of isoleucine. This enzyme is capable of binding to other peptides, such
Johannes Zschocke
Journal of inherited metabolic disease, 35(1), 81-89 (2011-12-01)
The HSD17B10 gene is located on chromosome Xp11.2 and codes for a multifunctional protein called 17β-hydroxysteroid dehydrogenase type 10 (HSD10). This protein catalyzes the 2-methyl-3-hydroxybutyryl-CoA dehydrogenation (MHBD) reaction in isoleucine metabolism and is an essential component of mitochondrial RNase P
Naeem Shafqat et al.
The Biochemical journal, 376(Pt 1), 49-60 (2003-08-15)
17beta-hydroxysteroid dehydrogenases (17beta-HSDs) catalyse the conversion of 17beta-OH (-hydroxy)/17-oxo groups of steroids, and are essential in mammalian hormone physiology. At present, eleven 17beta-HSD isoforms have been defined in mammals, with different tissue-expression and substrate-conversion patterns. We analysed 17beta-HSD type 10
Maria I G Lopez Sanchez et al.
Molecular endocrinology (Baltimore, Md.), 29(1), 14-27 (2014-11-07)
Estrogens, in particular 17β-estradiol, are well-known regulators of essential cellular functions; however, discrepancies remain over the mechanisms by which they act on mitochondria. Here we propose a novel mechanism for the direct regulation of mitochondrial gene expression by estrogen under
Johann Holzmann et al.
Cell, 135(3), 462-474 (2008-11-06)
tRNAs are synthesized as immature precursors, and on their way to functional maturity, extra nucleotides at their 5' ends are removed by an endonuclease called RNase P. All RNase P enzymes characterized so far are composed of an RNA plus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service