Skip to Content
Merck
All Photos(1)

Documents

H9017

Sigma-Aldrich

Heparin disaccharide II-H disodium salt

Synonym(s):

α-ΔUA-[1→4]-GlcN-6S

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C12H17NO13SNa2
CAS Number:
Molecular Weight:
461.31
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98%

form

powder

technique(s)

HPLC: suitable

storage temp.

−20°C

SMILES string

[O-]C(C1=C[C@H](O)[C@@H](O)[C@H](O[C@@]([C@H](O)COS([O-])(=O)=O)([H])[C@H](O)[C@H](C([H])=O)N)O1)=O.[Na+].[Na+]

InChI

1S/C12H19NO13S.2Na/c13-4(2-14)8(17)10(6(16)3-24-27(21,22)23)26-12-9(18)5(15)1-7(25-12)11(19)20;;/h1-2,4-6,8-10,12,15-18H,3,13H2,(H,19,20)(H,21,22,23);;/q;2*+1/p-2/t4-,5-,6+,8+,9+,10+,12-;;/m0../s1

InChI key

FOAZUBKCMDYJKE-HSLHGPCSSA-L

Biochem/physiol Actions

Products of the digestion of heparin and heparan sulfate by various heparinases

Other Notes

In the following listings, δUA = 4-deoxy-L-threo-hex-4-enopyranosyluronic acid; GlcN = D-glucosamine; Ac = Acetyl; NS, 2S, 6S, = N-sulfo, 2-sulfate and 6-sulfate respectively.
To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service