Skip to Content
Merck
All Photos(1)

Documents

H1879

Sigma-Aldrich

8-Hydroxyquinoline β-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17NO6
CAS Number:
Molecular Weight:
307.30
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

storage temp.

−20°C

SMILES string

OCC1OC(Oc2cccc3cccnc23)C(O)C(O)C1O

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Aranda et al.
Comptes rendus des seances de la Societe de biologie et de ses filiales, 176(1), 39-44 (1982-01-01)
A short-term method for detecting potential environmental carcinogens is described using in vitro transformation assay of human epithelial-like amniotic fluid cells. According to Styles, after a short exposition to the carcinogen, only transformed cells grow and form large colonies when
A L James et al.
Zentralblatt fur Bakteriologie, Mikrobiologie, und Hygiene. Series A, Medical microbiology, infectious diseases, virology, parasitology, 267(2), 188-193 (1987-12-01)
8-Hydroxyquinoline-beta-D-glucoside has been shown to be a useful alternative substrate for the differentiation of members of the family Enterobacteriaceae based on the presence or absence of beta-glucosidase. In an identification protocol based on multipoint-inoculated plates 8-hydroxyquinoline-beta-D-glucoside when incorporated into an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service