Skip to Content
Merck
All Photos(1)

Key Documents

GE17-1313-01

SDS

Cytiva 17-1313-01, pack of 100 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105323
NACRES:
NA.25

form

solid

feature

wetted part: no

packaging

pack of 100 g

manufacturer/tradename

Cytiva 17-1313-01

color

White to yellowish

pH range

6.5-8.5

solubility

acetone: soluble (Very slightly soluble in acetone)
cold water: soluble (Partially soluble in cold water)
diethyl ether: soluble (Insoluble in diethyl ether)
water and methanol: soluble

storage temp.

room temp

General description

SDS is an ionic detergent.

Application

Used to solubilize proteins.

Features and Benefits

  • The PlusOne SDS reagent is carefully controlled for carbon chain length and overall quality.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Storage Class Code

4.1B - Flammable solid hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

This page discusses column packing and preparation techniques for reverse phase chromatography.

Column, media, and sample preparation for Hydrophobic Interaction Chromatography (HIC) detailed with Cytiva media.

Protocols

This page discusses various aspects of sample preparation for chromatographic purification.

This page describes typical rehydration solutions for use in gel electrophoresis with Cytiva products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service