Skip to Content
Merck
All Photos(1)

Key Documents

F7804

Sigma-Aldrich

Fascaplysin chloride hydrate

≥98% (HPLC), solid

Synonym(s):

12,13-Dihydro-13-oxopyrido[1,2-a:3,4-b′]diindol-5-ium chloride, Fascaplysine hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H11ClN2O · xH2O
CAS Number:
Molecular Weight:
306.75 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

solid

color

brick red

solubility

H2O: <2 mg/mL
DMSO: ~20 mg/mL

storage temp.

2-8°C

SMILES string

[Cl-].[H]O[H].O=C1c2ccccc2-[n+]3ccc4c([nH]c5ccccc45)c13

InChI

1S/C18H10N2O.ClH.H2O/c21-18-13-6-2-4-8-15(13)20-10-9-12-11-5-1-3-7-14(11)19-16(12)17(18)20;;/h1-10H;1H;1H2

InChI key

WPIOPGXRERNYSI-UHFFFAOYSA-N

Biochem/physiol Actions

Originally isolated from the marine sponge Thorectandra sp., fascaplysin chloride is a selective, potent inhibitor of cyclin dependent kinase 4/cyclin D1 (IC50 = 0.35 μM). It is significantly less selective for Cdk6/D1 (IC50 = 3.4 μM) and not selective for the other Cdks and tyrosine kinases.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sachin Mahale et al.
Bioorganic chemistry, 34(5), 287-297 (2006-08-15)
Tryptamine derivatives, a new structural class of cyclin dependent kinase 4 inhibitors, have been identified during extensive biological screening of synthetic molecules. The molecules were synthesized based on the structure of fascaplysin, which is not only a specific inhibitor of
R Soni et al.
Biochemical and biophysical research communications, 275(3), 877-884 (2000-09-07)
Small chemical molecules that interfere with biological proteins could be useful for gaining insight into the complex biochemical processes in mammalian cells. Cdk4 is a key protein whose activity is required not only for emergence of cells from quiescence but
A M Popov et al.
Antibiotiki i khimioterapiia = Antibiotics and chemoterapy [sic], 36(1), 12-14 (1991-01-01)
Various aspects of the physiological activity of fascaplysine, a pigment from tropic sea sponges, were studied. One of the fragments of the chemical structure of the pigment is the indole ring. Ehrlich tumor cells, murine lymphocytes and erythrocytes were used
Carine Aubry et al.
Chemical communications (Cambridge, England), (15)(15), 1696-1697 (2004-07-28)
The first biologically active non-planar analogues of the toxic anti-cancer agent, fascaplysin, have been produced; we present the design, synthesis and biological activity of three tryptamine derivatives.
Nicole M R Schmitz et al.
The American journal of pathology, 169(3), 1074-1079 (2006-08-29)
Cyclin-dependent kinases (CDKs) successively phosphorylate the retinoblastoma protein (RB) at the restriction point in G1 phase. Hyperphosphorylation results in functional inactivation of RB, activation of the E2F transcriptional program, and entry of cells into S phase. RB unphosphorylated at serine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service