Skip to Content
Merck
All Photos(1)

Documents

F113

Sigma-Aldrich

R(−)-Fenfluramine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16F3N · HCl
CAS Number:
Molecular Weight:
267.72
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

drug control

USDEA Schedule IV

SMILES string

Cl.CCN[C@H](C)Cc1cccc(c1)C(F)(F)F

InChI

1S/C12H16F3N.ClH/c1-3-16-9(2)7-10-5-4-6-11(8-10)12(13,14)15;/h4-6,8-9,16H,3,7H2,1-2H3;1H/t9-;/m1./s1

InChI key

ZXKXJHAOUFHNAS-SBSPUUFOSA-N

Biochem/physiol Actions

Neurotoxic on prolonged administration or at high dosage; releases serotonin from axon terminals by a nonexocytotic mechanism; anorectic.
Less active enantiomer

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent studies in the clinical pharmacology of anorectic drugs.
Silverstone, et al.
Recent Adv. Obes. Res., 5, 285-285 (1986)
Can benfluorex induce congenital malformations?
I Lacroix et al.
Diabetes & metabolism, 38(4), 373-374 (2012-05-09)
Prescrire international, 22(135), 46-46 (2013-02-28)
Women were given appetite suppressants during pregnancy.
In the wake of the Mediator scandal: some progress in France, but apathy at the European level.
Prescrire international, 21(126), 110-110 (2012-04-21)
Mediator scandal engulfs French compensation body.
Asher Mullard
Lancet (London, England), 381(9880), 1803-1803 (2013-05-30)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service