Skip to Content
Merck
All Photos(1)

Key Documents

EHU045551

Sigma-Aldrich

MISSION® esiRNA

targeting human STK39

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCCACCCAATGCTAATGAAGACTACAGAGAAGCTTCTTCTTGTGCCGTGAACCTCGTTTTGAGATTAAGAAACTCCAGAAAGGAACTTAATGACATACGATTTGAGTTTACTCCAGGAAGAGATACAGCAGATGGTGTATCTCAGGAGCTCTTCTCTGCTGGCTTGGTGGATGGTCACGATGTAGTTATAGTGGCTGCTAATTTACAGAAGATTGTAGATGATCCCAAAGCTTTAAAAACATTGACATTTAAGTTGGCTTCTGGCTGTGATGGGTCGGAGATTCCTGATGAAGTGAAGCTGATTGGGTTTGCTCAGTTGAGTGTCAGCTGATGTATGTCCCTTGATGTCACCCTGATCTGTCATGCCCCACCGCCACCCCTACTCCCTTCAACCCTCCCTCTTTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ye Bi et al.
Frontiers in physiology, 11, 638-638 (2020-07-28)
SPS1-related proline/alanine-rich kinase (SPAK) plays important roles in regulating the function of numerous ion channels and transporters. With-no-lysine (WNK) kinase phosphorylates SPAK kinase to active the SPAK signaling pathway. Our previous studies indicated that WNK kinases regulate the activity of
Chih-Hao Shen et al.
BMC pulmonary medicine, 21(1), 58-58 (2021-02-17)
Hyperoxia downregulates the tight junction (TJ) proteins of the alveolar epithelium and leads to barrier dysfunction. Previous study has showed that STE20/SPS1-related proline/alanine-rich kinase (SPAK) interferes with the intestinal barrier function in mice. The aim of the present study is
Ken Yamada et al.
ACS chemical biology, 11(12), 3338-3346 (2016-10-08)
Protein kinases are known for their highly conserved adenosine triphosphate (ATP)-binding site, rendering the discovery of selective inhibitors a major challenge. In theory, allosteric inhibitors can achieve high selectivity by targeting less conserved regions of the kinases, often with an
Xiuyan Feng et al.
American journal of physiology. Renal physiology, 308(10), F1119-F1127 (2015-03-13)
Thiazide-sensitive sodium chloride cotransporter (NCC) plays an important role in maintaining blood pressure. Aldosterone is known to modulate NCC abundance. Previous studies reported that dietary salts modulated NCC abundance through either WNK4 [with no lysine (k) kinase 4]-SPAK (Ste20-related proline

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service