U-87 MG Cell Line human
89081402, human brain (glioblastoma astrocytoma), Epithelial-like
Synonym(s):
U-87MG Cells, U87 Cells, U87MG Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product name
U-87 MG Cell Line human, from human brain(glioblastoma astrocytoma), 89081402
biological source
human brain (glioblastoma astrocytoma)
growth mode
Adherent
karyotype
2n = 46
morphology
Epithelial-like
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human glioblastoma astrocytoma
Cell Line Description
Derived from a malignant glioma from a female patient by explant technique. It is reported to produce a malignant tumour consistent with glioblastoma in nude mice.
Application
Neural studies
U-87 MG cell line has been used to study the effects of the lysosomal destabilizing drug Siramesine on glioblastoma. It has also been used to study calcium-independent disruption of microtubule dynamics by nanosecond pulsed electric fields.
DNA Profile
STR-PCR Data: Amelogenin: X
CSF1PO: 10,11
D13S317: 8,11
D16S539: 12
D5S818: 11,12
D7S820: 8,9
THO1: 9.3
TPOX: 8
vWA: 15,17
CSF1PO: 10,11
D13S317: 8,11
D16S539: 12
D5S818: 11,12
D7S820: 8,9
THO1: 9.3
TPOX: 8
vWA: 15,17
Culture Medium
EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 1mM Sodium Pyruvate (NaP) + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service