B8429
Anti-Bax antibody, Mouse monoclonal
clone 6A7, purified from hybridoma cell culture
Synonym(s):
Anti-BCL2L4
About This Item
Recommended Products
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
6A7, monoclonal
form
buffered aqueous solution
mol wt
antigen 21 kDa (and possibly a dimer of 42 kDa)
species reactivity
mouse, rat, human
concentration
2 mg/mL
technique(s)
immunoprecipitation (IP): suitable
microarray: suitable
western blot: 10-60 μg/mL using extract of human breast adenocarcinoma MCF-7 cells, activated with dexamethasone
isotype
IgG1
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... BAX(581)
mouse ... Bax(12028)
rat ... Bax(24887)
Looking for similar products? Visit Product Comparison Guide
General description
Immunogen
Application
Biochem/physiol Actions
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
recommended
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service