Skip to Content
Merck
All Photos(1)

Documents

AC-0338A

Sigma-Aldrich

PAX8 (EP331) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

antibody form

Ig fraction of antiserum

clone

monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:200

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

General description

Paired box protein Pax-8 (PAX8) is a member of the PAX family of transcription factors. It is essential for organogenesis during embryonic development of the kidney, thyroid and paramesonephric ducts, giving rise to the urogenital organs including the seminal vesicles, vas deferens, ureter, uterus, and fallopian tubes. Although PAX8 is crucial for organogenesis, sustained expression in the normal kidney, thyroid, breast, seminal vesicles, ovarian, cervical, and endometrial epithelia suggest a role for homeostatic control of mature tissues. Immunohistochemical staining revealed almost exclusive nuclear localization, with low or negligible cytoplasmic expression. Due to the restrictive expression in normal tissues, PAX8 has been suggested to be a sensitive and specific marker for both primary and metastatic tumors derived from the above mentioned organs and tissues. PAX8 is expressed in renal cell carcinoma of clear cell, papillary and chromophobe subtypes. It is also found in endometrioid carcinoma, serous carcinoma, and thyroid carcinomas. However, PAX8 expression in lymphoid tissue has been controversial. The N-terminal polyclonal PAX8 antibody was found to cross-react with PAX5; previous data suggesting the use of PAX8 for diagnosis of malignant lymphomas should be re-evaluated. The cause may be due to a substantial sequence homology in the N-terminus of PAX8 and PAX5 (70%).

Quality


IVD

IVD

IVD

RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and &< 0.1% Sodium Azide

Other Notes

For a copy of the IFU and CofA contact IVDorder@ABCAM.com

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service