Skip to Content
Merck
All Photos(1)

Key Documents

A5156

Sigma-Aldrich

Actinomycin D–Mannitol

lyophilized powder

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$121.00

$121.00


Estimated to ship onApril 21, 2025


Request a Bulk Order

Select a Size

Change View
1 VIAL
$121.00

About This Item

MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

$121.00


Estimated to ship onApril 21, 2025


Request a Bulk Order

biological source

Streptomyces sp.

Quality Level

form

lyophilized powder

color

yellow to orange

solubility

H2O: 20 mg/mL

antibiotic activity spectrum

neoplastics

Mode of action

DNA synthesis | interferes

storage temp.

2-8°C

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.CC(C)[C@H]1NC(=O)[C@@H](NC(=O)c2ccc(C)c3OC4=CC(=O)C(N)=C(C(=O)N[C@H]5[C@@H](C)OC(=O)[C@H](C(C)C)N(C)C(=O)CN(C)C(=O)[C@@H]6CCCN6C(=O)[C@H](NC5=O)C(C)C)C4=Nc23)[C@@H](C)OC(=O)[C@H](C(C)C)N(C)C(=O)CN(C)C(=O)[C@@H]7CCCN7C1=O

InChI

1S/C61H84N12O16.C6H14O6/c1-27(2)43-58(83)72-22-16-18-35(72)56(81)68(12)25-39(75)70(14)49(29(5)6)60(85)87-32(10)45(54(79)64-43)66-52(77)34-21-20-31(9)51-47(34)63-48-38(89-51)24-37(74)42(62)41(48)53(78)67-46-33(11)88-61(86)50(30(7)8)71(15)40(76)26-69(13)57(82)36-19-17-23-73(36)59(84)44(28(3)4)65-55(46)80;7-1-3(9)5(11)6(12)4(10)2-8/h20-21,24,27-30,32-33,35-36,43-46,49-50H,16-19,22-23,25-26,62H2,1-15H3,(H,64,79)(H,65,80)(H,66,77)(H,67,78);3-12H,1-2H2/t32-,33-,35+,36+,43-,44-,45+,46+,49+,50+;3-,4-,5-,6-/m11/s1

InChI key

AURFRHZIDSMTRK-CSXPNASWSA-N

General description

Chemical structure: peptide

Application

Actinomycin D is used as a selection agent and is used in banding techniques to differentiate between different regions of chromosomes. Actinomycin induces apoptosis and is used as a potent antitumor agent [1].

Biochem/physiol Actions

An antineoplastic antibiotic that inhibits cell proliferation by forming a stable complex with DNA and blocking the movement of RNA polymerase which interferes with DNA-dependent RNA synthesis. Induces apoptosis. Potent antitumor agent. For cell culture applications, actinomycin D is used as a selection agent and in banding techniques to differentiate between different regions of chromosomes.
Actinomycin D is an antineoplastic antibiotic that inhibits cell proliferation by forming a stable complex with DNA and blocking the movement of RNA polymerase which interferes with DNA-dependent RNA synthesis. Induces apoptosis. Potent antitumor agent.
An antineoplastic antibiotic that inhibits cell proliferation by forming a stable complex with DNA and blocking the movement of RNA polymerase which interferes with DNA-dependent RNA synthesis. Induces apoptosis. Potent antitumor agent. For cell culture applications, actinomycin D is used as a selection agent and is used in banding techniques to differentiate between different regions of chromosomes.

Components

1 mg actinomycin D and 49 mg mannitol per vial

Other Notes

The powder is hygroscopic and sensitive to light.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral - Carc. 2 - Resp. Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

G S Ridge et al.
Nucleic acids research, 22(24), 5241-5246 (1994-12-11)
The antitumour antibiotic actinomycin D normally binds to DNA by intercalation at sequences containing the CpG step, but in the presence of daunomycin it has been reported to interact with poly(dA-dT). This observation has neither been confirmed nor explained. Here
Haishan Xu et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 16(17), 4392-4400 (2010-08-25)
ABT-737, which blocks the function of Bcl-2 and Bcl-X(L) but not Mcl-1, has shown single-agent activity in preclinical models of small cell lung cancer (SCLC). Elevated expression of Mcl-1 induces resistance to ABT-737 in SCLC. Based on the short half-life
Louis-Marie Bloyet et al.
Journal of virology, 94(13) (2020-04-24)
Nonsegmented negative-strand (NNS) RNA viruses possess a ribonucleoprotein template in which the genomic RNA is sequestered within a homopolymer of nucleocapsid protein (N). The viral RNA-dependent RNA polymerase (RdRP) resides within an approximately 250-kDa large protein (L), along with unconventional
William J Neidermyer et al.
PLoS pathogens, 15(6), e1007875-e1007875 (2019-06-22)
Infection of mammalian cells with vesicular stomatitis virus (VSV) results in the inhibition of cellular translation while viral translation proceeds efficiently. VSV RNA synthesis occurs entirely within the cytoplasm, where during transcription the viral polymerase produces 5 mRNAs that are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service