Skip to Content
Merck
All Photos(1)

Key Documents

A3846

Sigma-Aldrich

AL-8810

≥98% (HPLC), solid

Synonym(s):

(5Z, 13E)-(9S,11S,15R)-9,15,Dihydroxy-11-fluoro-15-(2-indanyl)-16,17,18,19,20,pentanor-5,13-prostadienoic acid, [1R-[1α(Z),2β(1E,3S*),3β,5α]]-7-[3-Fluoro-5-hydroxy-2-[3-hydroxy-4-[2-(2,3-dihydro-1H-indenyl)]-1-butenyl]cyclolpentyl]-5-heptenoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$688.00

$688.00


Estimated to ship onApril 20, 2025



Select a Size

Change View
5 MG
$688.00

About This Item

Empirical Formula (Hill Notation):
C24H31FO4
CAS Number:
Molecular Weight:
402.50
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$688.00


Estimated to ship onApril 20, 2025


Quality Level

Assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

DMSO: soluble 10 mg/mL
ethanol: soluble

storage temp.

2-8°C

SMILES string

O[C@H]1C[C@H](F)[C@H](\C=C\[C@H](O)C2Cc3ccccc3C2)[C@H]1C\C=C/CCCC(O)=O

InChI

1S/C24H31FO4/c25-21-15-23(27)20(9-3-1-2-4-10-24(28)29)19(21)11-12-22(26)18-13-16-7-5-6-8-17(16)14-18/h1,3,5-8,11-12,18-23,26-27H,2,4,9-10,13-15H2,(H,28,29)/b3-1-,12-11+/t19-,20-,21+,22+,23+/m1/s1

InChI key

WTYSXBKKVNOOIX-JTGCGUAKSA-N

Gene Information

human ... PTGFR(5737)

Biochem/physiol Actions

AL-8810 is a novel prostaglandin F analog; selective FP prostanoid receptor antagonist.

Legal Information

Sold under license from Alcon.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dedmer Schaafsma et al.
Respiratory research, 6, 85-85 (2005-07-29)
In addition to their proliferative and differentiating effects, several growth factors are capable of inducing a sustained airway smooth muscle (ASM) contraction. These contractile effects were previously found to be dependent on Rho-kinase and have also been associated with the
Evanthia Nikolopoulou et al.
Journal of lipid research, 55(12), 2479-2490 (2014-10-18)
Arachidonic acid (AA) is a major PUFA that has been implicated in the regulation of adipogenesis. We examined the effect of a short exposure to AA at different stages of 3T3-L1 adipocyte differentiation. AA caused the upregulation of fatty acid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service