Skip to Content
Merck
All Photos(1)

Key Documents

Z734438

ThermalSeal RTS Sealing Films

for qPCR, storage & crystallization, non-sterile

Synonym(s):

qPCR plate seal, storage plate seal

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$527.00

$527.00


Estimated to ship onMarch 15, 2025



Select a Size

Change View
100 EA
$527.00

About This Item

UNSPSC Code:
41122410
NACRES:
NB.22

$527.00


Estimated to ship onMarch 15, 2025


material

polyolefin film
silicone/adhesive

sterility

non-sterile

packaging

pack of 100 ea

manufacturer/tradename

Excel Scientific TSS-RTQ-100

technique(s)

PCR: suitable
qPCR: suitable

L × W

76.2 mm × 133.4 mm

W × L

7.62 cm × 13.34 cm

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

ThermalSeal RTS sealing film is a 50 μm thick polyolefin with 50 μm inert encapsulated silicone adhesive. They are optically clear, with low autofluorescence, and are especially suited for real-time qPCR, storage, and protein crystallization applications. The encapsulated silicone adhesive is non-tacky until pressed against the sealing surface, at which time adhesive is released only in sealing areas to form the strongest available heat-resistant seal around each well on the plate. Adhesive on non-sealing areas of the film, such as directly over sample wells, remains encapsulated and inert.

ThermalSeal RTS films are sized to fit within the edges of raised-rim 96-well plates. Their consistent high optical clarity makes possible reproducible, reliable, and consistent DNA amplification measurements and crystal detection. Two end tabs assist in positioning the film on the plate, and the non-tacky adhesive surface simplifies handling. Easy removal of the end tabs at perforated boundaries prevents lifting and higher evaporation rates that can occur with films that overlap the plate rim.

Dimensions 76.2 by 133.4 mm. With end tabs removed, length is 113.0 mm.

  • High optical clarity
  • Minimal to no autofluorescence
  • Chemically inert; no extractables except at extreme pH
  • DMSO resistant for HTS
  • Heat resistant, recommended for temperatures from
  • -70°C to +100°C
  • Certified DNase, RNase, and Nucleic Acid free
  • Fit within raised plate rim to prevent loss of seal due to film lifting
  • Silicone adhesive forms the strongest available seal for evaporation prevention
  • Non-tacky adhesive layer simplifies handling of film prior to sealing

Legal Information

ThermalSeal is a registered trademark of Excel Scientific, Inc.
ThermalSeal RTS is a trademark of Excel Scientific, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Primer Concentration Optimization Protocol creates reaction matrix for testing primer concentrations against various partners.

Reverse transcription analyzes mRNA gene expression, facing challenges like transcript half-life differences and temporal patterns.

Perform reverse transcription (RT) using a reverse transcriptase enzyme and dNTPs. Use total RNA or a gene-specific approach so that only the RNA of interest is converted to cDNA.

Standard qPCR protocol with up to four detection probes at specified concentrations, simplifying reaction setup with LuminoCt ReadyMix.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service