Z374903
PCR multiwell plates
96 well plate for PCR
Synonym(s):
96 mulitwell PCR plate, 96 well PCR microplate, 96 well PCR plate, 96 well microtiter plate
About This Item
Recommended Products
material
colorless polypropylene
colorless wells
sterility
non-sterile
feature
plate format: 96 well standard
packaging
case of 25 ea
manufacturer/tradename
Sorenson 2633
technique(s)
PCR: suitable
size
96 well
well maximum volume
300 μm
working volume
200 μL
suitability
suitable for (PCR, RT-PCR or DNA purification applications)
application(s)
agriculture
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Virgin polypropylene
- Fully autoclavable
- Certified DNase- and RNase-free
- Wells have thin walls for rapid temperature equilibration and reduced cycle time
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
Reverse transcription (RT) is the process of converting RNA to cDNA using a reverse transcriptase enzyme and dNTPs.
Hot Start dNTPs block DNA polymerase until heat activation, enhancing PCR specificity.
Primer Concentration Optimization Protocol creates reaction matrix for testing primer concentrations against various partners.
Reverse transcription analyzes mRNA gene expression, facing challenges like transcript half-life differences and temporal patterns.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service