Skip to Content
Merck
All Photos(1)

Key Documents

SIAL0828

Sigma-Aldrich

Sigma® centrifuge tubes

capacity 50 mL, pack of 20 × 25 ea racked, sterile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122804
NACRES:
NB.22

Pricing and availability is not currently available.

material

polypropylene

description

marking spot

sterility

sterile

feature

RCF 17000 × g
disposable
graduated
graduations 5 ‑ 50 mL (graduation interval: 2.5mL)

packaging

pack of 20 × 25 ea racked

O.D. × L

29.1 mm × 114.4 mm

capacity

50 mL

color

natural

Looking for similar products? Visit Product Comparison Guide

General description

Sigma disposable 15 mL centrifuge tubes are available as clear polyethylene terephthalate (PET) tubes with a traditional plug seal cap or polypropylene (PP) tubes with either plug seal caps or the new easy-on/easy-off top offering advanced ergonomics with its wider knurls and roll-over edge design. The Sigma centrifuge tubes are available as polypropylene tubes either racked or bulk packed.

  • Sterile, certified nonpyrogenic and DNase-/RNase-free
  • Black printed graduations and a large white marking spot
  • Maximum RCF for PET tubes is 3,600xg

Legal Information

Sigma is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

3D Spheroid BME Cell Invasion Assay utilizes specialized plates and ECM for cell aggregation and spheroid formation in vitro.

3-D Culture Matrix™ Collagen I facilitates cellular growth and differentiation in vitro, offering versatile applications in biomedical research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service