Skip to Content
Merck
All Photos(1)

Key Documents

S7026

Sigma-Aldrich

Sodium thiosulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S2O3
CAS Number:
Molecular Weight:
158.11
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

98% (NLT)

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=S

InChI

1S/2Na.H2O3S2/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

AKHNMLFCWUSKQB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Used with silver nitrate to produce a silver thiosulfate solution (STS) containing the ethylene inhibitor ion [Ag(S2O3)2]-3.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Michael C Reade et al.
Emergency medicine Australasia : EMA, 24(3), 225-238 (2012-06-08)
Cyanide poisoning is uncommon, but generates interest because of the presumed utility of an antidote immediately available in those areas with a high risk of cyanide exposure. As part of its regular review of guidelines, the Australian Resuscitation Council conducted
Melani Sooriyaarachchi et al.
Metallomics : integrated biometal science, 4(9), 960-967 (2012-07-31)
The anticancer drug cis-platin (CP) is widely used to treat patients, but it is also associated with significant side effects, including nephrotoxicity. Given that this metallodrug is intravenously (iv) administered, its biotransformations in the bloodstream are likely to be involved
S Van Haute et al.
International journal of food microbiology, 208, 102-113 (2015-06-13)
A methodology to i) assess the feasibility of water disinfection in fresh-cut leafy greens wash water and ii) to compare the disinfectant efficiency of water disinfectants was defined and applied for a combination of peracetic acid (PAA) and lactic acid
Alan H Hall et al.
Annals of emergency medicine, 49(6), 806-813 (2006-11-14)
Cyanide poisoning must be seriously considered in victims of smoke inhalation from enclosed space fires; it is also a credible terrorism threat agent. The treatment of cyanide poisoning is empiric because laboratory confirmation can take hours or days. Empiric treatment
Edward A Levine et al.
Journal of the American College of Surgeons, 218(4), 573-585 (2014-02-05)
Peritoneal dissemination of abdominal malignancy (carcinomatosis) has a clinical course marked by bowel obstruction and death; it traditionally does not respond well to systemic therapy and has been approached with nihilism. To treat carcinomatosis, we use cytoreductive surgery (CS) with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service