Skip to Content
Merck
All Photos(1)

Key Documents

S5381

Sigma-Aldrich

Stevia rebaudiana bertoni leaves

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Application

Stevia rebaudiana bertoni leaves may be used as a source of stevioside, an FDA approved nontoxic natural non-caloric sweetener. Sevioside, a diterpene glycoside, is use to research its biological properties such as its anti-diabetic, anti-inflammatory and anti-tumor activities.

Preparation Note

Air-dried leaves containing stevioside

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Paul et al.
Nutrition and cancer, 64(7), 1087-1094 (2012-10-16)
Stevioside is a diterpene glycoside found in the leaf of Stevia rebaudiana, a traditional oriental medicinal herb, which has been shown to have various biological and ethno-medicinal activities including antitumor activity. In this study, we investigated the effects of stevioside
Li Fengyang et al.
Inflammation, 35(5), 1669-1675 (2012-05-31)
Stevioside, a diterpene glycoside isolated from Stevia rebaudiana, has been reported to have anti-inflammatory properties. However, the underlying molecular mechanisms are not well understood. The objective of this study was to investigate the molecular mechanism of stevioside in modifying lipopolysaccharide
Indu Barwal et al.
Colloids and surfaces. B, Biointerfaces, 101, 510-516 (2012-10-02)
Stevioside (FDA approved nontoxic natural non-caloric sweetener) has been reported to have very good antidiabetic potential but its use as therapeutic drug is restricted in human due to its deprived intestinal absorption and poor bioavailability. We have nano-bioconjugated this molecule

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service