Skip to Content
Merck
All Photos(1)

Key Documents

R1386

Sigma-Aldrich

RNA Sample Loading Buffer

for NA electrophoresis, without ethidium bromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 VIALS
$151.00

$151.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
5 VIALS
$151.00

About This Item

EC Number:
MDL number:
UNSPSC Code:
12161700
NACRES:
NA.25

$151.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

grade

Molecular Biology
for molecular biology

Quality Level

form

solution

foreign activity

RNase, none detected

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

RNA loading buffer is used as a tracking dye during RNA electrophoresis. The dye has a slight negative charge and will migrate the same direction as RNA, allowing the user to monitor the progress of molecules moving through the gel. The rate of migration varies with gel composition. Dilute 1:3 to 1:6 with sample, heat to 65C for ten minutes and chill on ice before loading.

Application

RNA sample loading buffer is especially formulated for electrophoresis of RNA on formaldehyde-agarose gels with or without ethidium bromide. Ethidium bromide is not recommended for gel staining prior to Northern blot detection because the presence of ethidium bromide in agarose gels or the loading buffer can cause poor transfer efficiency.
Suitable for use with formaldehyde-agarose gels used in Northern blotting procedures.[1]

Components

Deionized formamide 62.5% (v/v), formaldehyde 1.14 M, bromphenol blue 200 μg/mL, xylene cyanole 200 μg/mL, MOPS-EDTA-sodium acetate at 1.25× working concentration.
RNA loading buffer contains 62.5% deionized formamide, 1.14M formaldehyde, 200 μg/ml bromphenol blue, 200 μg/ml xylene cyanole, in MOPS-EDTA-sodium acetate at 1.25x working concentration.

Quantity

Recommended usage: Add 1 volume sample to 2-5 volumes of sample loading buffer and mix well. The sample should be heated to 65 °C for 10 minutes, and then chilled on ice immediately before loading on the gel.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gene expression analysis of frontotemporal lobar degeneration of the motor neuron disease type with ubiquitinated inclusions.
Mishra M, et al.
Acta Neuropathologica, 114, 81-94 (2007)
Ahmad Kobiita et al.
Cell reports, 32(1), 107846-107846 (2020-07-09)
The ability of pancreatic β-cells to respond to increased demands for insulin during metabolic stress critically depends on proper ribosome homeostasis and function. Excessive and long-lasting stimulation of insulin secretion can elicit endoplasmic reticulum (ER) stress, unfolded protein response, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service