PCR9604
GenElute™ 96 Well PCR Clean-Up Kit
4 x 96 well plate, sufficient for 4 96-well plate purifications
Synonym(s):
High Throughput Purification, PCR Purification, PCR Purification, PCR Purification Kit
About This Item
Recommended Products
usage
sufficient for 4 96-well plate purifications
quality
4 x 96 well plate
technique(s)
DNA purification: suitable
storage temp.
15-25°C
Looking for similar products? Visit Product Comparison Guide
General description
Once the PCR reaction is complete, the reaction volume is adjusted and Binding Solution is added. The samples are transferred to the Binding Plate where the PCR reaction is captured on the silica membrane. The bound PCR product is washed several times to remove primers, primer-dimers, salts, nucleotides, and polymerases. Finally the purified PCR product is eluted and ready for immediate use in downstream applications.
The purified PCR product is ready for a variety of downstream applications including sequencing, restriction digest, ligation, and microarray analysis.
Application
Features and Benefits
- Complete removal of primers and primer-dimers
- Innovative wash plate minimizes risk of cross-contamination
- Suitable for processing under vacuum and centrifugation
- Time-saving parallel clean-up of PCR products
Principle
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C
Supplementary Hazards
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service