Skip to Content
Merck
All Photos(1)

Documents

M8403

Sigma-Aldrich

McCoy′s 5A Medium

With sodium bicarbonate, without ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

5A basal medium

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

product name

McCoy′s 5A Medium, Modified, with sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

L-glutamine: no
NaHCO3: 2.2 g/L
glucose: 3.0 g/L
phenol red: 0.011 g/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

McCoy′s 5A Medium is suitable for the primary culture of cells from a wide range of tissue. It is recommended for the production of viruses in primary cell cultures.

Application

McCoy′s 5A Medium has been used to maintain:

  • G-361 melanoma cells;
  • 87-MG cells
  • human colon carcinoma cell line HCT116

Biochem/physiol Actions

McCoy′s 5A Medium has been originally developed to support liver tumor cells by modification of the amino acids found in BME. This formulation has also been used to support the growth of primary cultures of bone marrow, skin, gingiva, kidney, omentum, adrenals, lung, spleen, rat embryos, and other cell types.

Reconstitution

Supplement with 0.22 g/L L-glutamine.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katsuhiro Uzawa et al.
Scientific reports, 4, 5433-5433 (2014-06-26)
No definitive therapy exists to treat human metastatic tumors. We reported previously that down-regulation of Lin-7C is essential for metastasis of human squamous cell carcinomas (hSCCs). In this study, we investigated the chemical restoration of Lin-7C expression and demonstrated its
Seungman Park et al.
Scientific reports, 9(1), 13286-13286 (2019-09-19)
We present a high-throughput microfluidics technique facilitating in situ measurements of cell mechanics parameters at the dorsal side of the cell, including molecular binding strengths, local traction forces, and viscoelastic properties. By adjusting the flow rate, the force magnitude exerted
Claudia De Fusco et al.
Journal of medicinal chemistry, 64(10), 6814-6826 (2021-04-27)
MAT2a is a methionine adenosyltransferase that synthesizes the essential metabolite S-adenosylmethionine (SAM) from methionine and ATP. Tumors bearing the co-deletion of p16 and MTAP genes have been shown to be sensitive to MAT2a inhibition, making it an attractive target for
Huw D Summers et al.
ACS nano, 7(7), 6129-6137 (2013-06-19)
Assessing dose in nanoparticle-cell interactions is inherently difficult due to a complex multiplicity of possible mechanisms and metrics controlling particle uptake. The fundamental unit of nanoparticle dose is the number of particles internalized per cell; we show that this can
A-P Sappino et al.
Oncogenesis, 1, e7-e7 (2012-01-01)
The p53 tumor-suppressor protein has a key role in the induction of cellular senescence, an important barrier to cancer development. However, very little is known about the physiological mediators of cellular senescence induced by p53. CEACAM1 is an immunoglobulin superfamily

Articles

McCoy's 5A Modified Media Formulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service