Skip to Content
Merck
All Photos(5)

Documents

E5038

Sigma-Aldrich

Plant/Fungi DNA Isolation Kit

sufficient for 50 purifications

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501

usage

sufficient for 50 purifications

Quality Level

General description

Norgen Biotek′s Plant/Fungi DNA Isolation Kit provides a rapid method for the isolation and purification of total DNA from a wide range of plant and filamentous fungal species. Total DNA, including genomic DNA, mitochondrial DNA and chloroplast DNA can be purified from fresh or frozen plant tissues, plant cells or filamentous fungi samples using this kit.

Application

Norgen′s Plant/Fungi DNA Isolation Kit purifies high quality total DNA that can be used in a number of downstream applications, including:
  • qPCR
  • Southern blotting
  • SNP analysis
  • Sequencing
Plant/Fungi DNA Isolation Kit has been used for rapid isolation of total DNA from plants and fungi.

Other Notes

Storage Conditions and Product Stability
All solutions should be kept tightly sealed and stored at room temperature. These reagents should remain stable for at least 1 year in their unopened containers.

related product

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chin Hong Neoh et al.
Environmental science and pollution research international, 20(5), 2912-2923 (2012-10-12)
The conventional treatment process of palm oil mill effluent (POME) produces a highly colored effluent. Colored compounds in POME cause reduction in photosynthetic activities, produce carcinogenic by-products in drinking water, chelate with metal ions, and are toxic to aquatic biota.
Distribution and occurrence of some pteridophytes in Gujarat: a new record for the state
Ronak N
Journal of Indian Botanical Society , 236-244 (2015)
Xuanqing Wang et al.
Journal of environmental management, 270, 110958-110958 (2020-07-30)
Composting is a cost-efficient method of transferring various unstable and complex organic matters into a stable and humus-like substance, during which various fungus play a critical role in the decomposition of organic matters. In this study, the rice straw and
J Yang et al.
Genetics and molecular research : GMR, 15(4) (2016-11-05)
Fusarium wilt (also known as Panama disease) is one of the most destructive banana diseases, and greatly hampers the global production of bananas. Consequently, it has been very detrimental to the Chinese banana industry. An infected plant is one of
Isolation and identification of Trichoderma harzianum from groundwater: An effective biosorbent for defluoridation of groundwater.
Koshle S
Journal of environmental biology / Academy of Environmental Biology, India, 37(1), 135-140 (2016)

Articles

Simple DNA/RNA purification methods aid genome analysis from various sources, enhancing research efficiency.

Protocols

The Plant RNA/DNA Purification Kit provides a rapid method for the isolation and purification of total RNA and DNA simultaneously from a single sample of plants.

The Plant/Fungi DNA Isolation Kit provides a rapid method for the isolation and purification of total DNA from a wide range of plant and fungi species.

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Related Content

Food processing practices are common in today's agricultural and food industries, and are often harsh on the genetic information of plants and plant products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service