Skip to Content
Merck
All Photos(1)

Key Documents

D9690

Sigma-Aldrich

β-Defensin 2 human

recombinant, expressed in E. coli, lyophilized powder (from 10 mM acetic acid)

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
$695.00

$695.00


Estimated to ship onApril 16, 2025



Select a Size

Change View
5 μG
$695.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

$695.00


Estimated to ship onApril 16, 2025


recombinant

expressed in E. coli

Quality Level

form

lyophilized powder (from 10 mM acetic acid)

mol wt

estimated mol wt 4.3 kDa

UniProt accession no.

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
fungi
viruses

Mode of action

cell wall synthesis | interferes

storage temp.

−20°C

Gene Information

human ... DEFB4(1673)

Amino Acid Sequence

Gly-Ile-Gly-Asp-Pro-Val-Thr-Cys-Leu-Lys-Ser-Gly-Ala-Ile-Cys-His-Pro-Val-Phe-Cys-Pro-Arg-Arg-Tyr-Lys-Gln-Ile-Gly-Thr-Cys-Gly-Leu-Pro-Gly-Thr-Lys-Cys-Cys-Lys-Lys-Pro

General description

Chemical structure: peptide

Biochem/physiol Actions

A small cationic peptide which demonstrates anti-microbial activity and is expressed in keratinocytes and the epithelia of many organs.
Anti-microbial expressed in keratinocytes and epithelia of many organs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Ribosomally synthesized antimicrobial peptides are a promising focus in antibiotic research amidst bacterial resistance and emerging infectious diseases.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service