Skip to Content
Merck
All Photos(1)

Key Documents

B4408

Sigma-Aldrich

Scienceware® biohazard disposal bags

W × H × thickness 24 in. (61 cm) × 36 in. (91 cm) × 0.00125 in., translucent, pack of 100

Synonym(s):

biohazard disposal bags

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$247.00

$247.00


Estimated to ship onApril 17, 2025



Select a Size

Change View
100 EA
$247.00

About This Item

UNSPSC Code:
24111500
NACRES:
NB.35

$247.00


Estimated to ship onApril 17, 2025


material

translucent

description

Imprinted in red with biohazard symbol and precautions in English, Spanish, French, and German.

packaging

pack of 100

manufacturer/tradename

Bel-Art F13162-0009

W × H × thickness

24 in. (61 cm) × 36 in. (91 cm) × 0.00125 in.

Looking for similar products? Visit Product Comparison Guide

Application

Scienceware® biohazard disposal bag is suitable for the disposal of cell culture flasks, culture plates, pipettes, petri dishes, multi-well cell culture plates, membrane filters and other biohazard wastes.

Features and Benefits

Scienceware® biohazard disposal bag provides the following features and benefits:
  • Made of polypropylene and can withstand high autoclave temperatures up to 135°C (275°F)
  • Cost-effective
  • Available as plain or non-printed or with biohazard-label and indicator patch
  • Sterilization indicator patch darkens during steam sterilization
  • Printed and non-printed bags can be used as trash can liners

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Available Fluorescent in situ hybridization (FISH) procedures, reagents and equipment.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service