Skip to Content
Merck
All Photos(1)

Key Documents

906H-0

Sigma-Aldrich

GCDFP-15 (23A3) + Mammaglobin Cocktail (304-1A5 & 31A5) Mouse/Rabbit Monoclonal Antibodies

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse
rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

23A3 + 304-1A5&31A5, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

bottle of 1.0 mL predilute (906H-07)
bottle of 7.0 mL predilute (906H-08)

concentration

0.01-1 μg/mL (predilute)

isotype

IgG1
IgG2a
IgG

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... PIP(5304)

General description

Anti-GCDFP-15, mouse monoclonal (23A3), and anti-mammaglobin, mouse monoclonal (304-1A5) and rabbit monoclonal (31A5), is an antibody cocktail. GCDFP-15 is a 15 kDa glycoprotein which is localized in the apocrine metaplastic epithelium lining breast cysts and in apocrine glands in the axilla, vulva, eyelid, and ear canal. Mammaglobin (10 kDa) is a breast-associated glycoprotein distantly related to secretoglobin family that includes human uteroglobin and lipophilin.

Quality


IVD

IVD

IVD

RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and format
Note: This requires a keycode which can be found on your packaging or product label.

Download the latest released IFU
Note: This IFU may not apply to your specific product lot.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuji Takeda et al.
Archives of pathology & laboratory medicine, 132(2), 239-243 (2008-02-07)
The lung is the most common site of metastasis during the natural history of malignant tumors. Breast carcinoma has a propensity for distant metastasis, and the lung and pleura are among the most common metastatic sites. Although it is often
C Cohen et al.
Archives of pathology & laboratory medicine, 117(3), 291-294 (1993-03-01)
Mammary Paget's disease has been said to result from epidermal spread by contiguity of primary intraductal carcinoma. To assess similar identity, we immunostained Paget's cells and underlying intraductal and/or invasive mammary carcinoma in 20 cases for cytokeratins, epithelial membrane antigen
Rohit Bhargava et al.
American journal of clinical pathology, 127(1), 103-113 (2006-12-06)
There are limited data that compare the usefulness of mammaglobin with gross cystic disease fluid protein-15 (GCDFP-15) in the identification of breast carcinomas. Whole tissue sections of 29 breast carcinomas with matched lymph node metastases and 63 breast carcinomas on
B Liegl et al.
Histopathology, 50(4), 439-447 (2007-04-24)
Mammary Paget's disease (MPD) and extramammary Paget's disease (EMPD) are rare neoplasms. The aim of this study was, by the use of immunohistochemistry, to derive further information about the cell(s) of origin, find a diagnostically useful immunohistochemical panel and investigate
Carmen Tornos et al.
The American journal of surgical pathology, 29(11), 1482-1489 (2005-10-15)
Metastatic breast carcinoma to the ovary is sometimes difficult to differentiate from primary ovarian carcinoma. This problem is often encountered in breast carcinoma patients who develop adnexal masses. ER and PR can be positive in a high percentage of breast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service