Skip to Content
Merck
All Photos(1)

Key Documents

80114

Sigma-Aldrich

1,2-Dimyristoyl-sn-glycero-3-phospho-L-serine sodium salt

≥70% (TLC)

Synonym(s):

(2S,8R)-2-Amino-5-hydroxy-11-oxo-8-[(1-oxotetradecyl)oxy]-4,6,10-trioxa-5-phosphatetracosanoic acid 5-oxide monosodium salt, 1,2-Ditetradecanoyl-sn-glycero-3-phosphoserine sodium salt, 1,2-Ditetradecanoyl-sn-glycero-3-phospho-L-serine sodium salt, 3-sn-Phosphatidyl-L-serine, dimyristoyl sodium salt, DMPS-Na, Dimyristoyl phosphatidylserine sodium salt, PS(14:0/14:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H65NNaO10P
CAS Number:
Molecular Weight:
701.84
MDL number:
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.85

biological source

synthetic

Assay

≥70% (TLC)

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

storage temp.

−20°C

SMILES string

[Na+].CCCCCCCCCCCCCC(=O)OC[C@H](COP([O-])(=O)OC[C@H](N)C(O)=O)OC(=O)CCCCCCCCCCCCC

InChI

1S/C34H66NO10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-32(36)42-27-30(28-43-46(40,41)44-29-31(35)34(38)39)45-33(37)26-24-22-20-18-16-14-12-10-8-6-4-2;/h30-31H,3-29,35H2,1-2H3,(H,38,39)(H,40,41);/q;+1/p-1/t30-,31+;/m1./s1

InChI key

QSHQBWBFNCFHLO-MFABWLECSA-M

Application

The effect of daidzein on cell membranes was modeled by studies with synthetic liposomes, which are extensively aggregated by daidzein. DMPS formed the model saturated-fatty-acid liposomes, which are aggregated below their main phase transition temperature, but not above it.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elżbieta Gryz et al.
International journal of molecular sciences, 20(12) (2019-06-27)
 Mono-saturated polyprenols (dolichols) have been found in almost all Eukaryotic cells, however, dolichols containing additional saturated bonds at the ω-end, have been identified in A.fumigatus and A.niger. Here we confirm using an LC-ESI-QTOF-MS analysis, that poly-saturated dolichols are abundant in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service