19474
Eriodictyol 7-O-β-D-glucopyranoside
≥99.0% (HPLC)
Synonym(s):
(2S)-Eriodictyol 7-O-β-D-glucopyranoside, 3′,4′,5,7-Tetrahydroxyflavanone 7-β -D-glucopyranoside, Eriodictyol 7-O-glucoside, Miscanthoside
About This Item
Recommended Products
biological source
plant leaves (Acer mono)
Assay
≥99.0% (HPLC)
form
powder
technique(s)
HPLC: suitable
mp
252 °C
storage temp.
2-8°C
SMILES string
OC[C@H]1O[C@@H](Oc2cc(O)c3C(=O)C[C@H](Oc3c2)c4ccc(O)c(O)c4)[C@H](O)[C@@H](O)[C@@H]1O
InChI
1S/C21H22O11/c22-7-16-18(27)19(28)20(29)21(32-16)30-9-4-12(25)17-13(26)6-14(31-15(17)5-9)8-1-2-10(23)11(24)3-8/h1-5,14,16,18-25,27-29H,6-7H2/t14-,16+,18+,19-,20+,21+/m0/s1
InChI key
RAFHNDRXYHOLSH-SFTVRKLSSA-N
General description
Biochem/physiol Actions
Packaging
Other Notes
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service