Skip to Content
Merck
All Photos(1)

Key Documents

PHR1370

Supelco

Benzyl Nicotinate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Benzyl nicotinate, Nicotinic acid benzyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$123.00

$123.00


Estimated to ship onApril 25, 2025


Request a Bulk Order

Select a Size

Change View
1 ML
$123.00

About This Item

Empirical Formula (Hill Notation):
C13H11NO2
CAS Number:
Molecular Weight:
213.23
Beilstein:
159169
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$123.00


Estimated to ship onApril 25, 2025


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

API family

benzyl nicotinate

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

177 °C/8 mmHg (lit.)

mp

24 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

O=C(OCc1ccccc1)c2cccnc2

InChI

1S/C13H11NO2/c15-13(12-7-4-8-14-9-12)16-10-11-5-2-1-3-6-11/h1-9H,10H2

InChI key

KVYGGMBOZFWZBQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Benzyl Nicotinate is a medication widely used as a vasodilator, that widens blood vessels. It basically oxygenates and flushes the skin′s capillaries.[1]

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3252 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

How to Be Inappropriate (2009)
C Albèr et al.
International journal of cosmetic science, 36(5), 412-418 (2014-05-03)
The mammalian skin is a barrier that effectively separates the water-rich interior of the body from the normally dryer exterior. Changes in the external conditions, for example ambient humidity, have been shown to affect the skin barrier properties. The prime

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service