Skip to Content
Merck
All Photos(1)

Documents

C3210

Sigma-Aldrich

Calcium sulfate dihydrate

BioXtra, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaSO4 · 2H2O
CAS Number:
Molecular Weight:
172.17
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

product line

BioXtra

Assay

≥98.0%

reaction suitability

reagent type: catalyst
core: calcium

impurities

≤0.005% P
≤0.02% Insoluble matter

anion traces

chloride (Cl-): ≤0.005%

cation traces

Al: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.001%
K: ≤0.005%
Mg: ≤0.02%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

O.O.[Ca++].[O-]S([O-])(=O)=O

InChI

1S/Ca.H2O4S.2H2O/c;1-5(2,3)4;;/h;(H2,1,2,3,4);2*1H2/q+2;;;/p-2

InChI key

PASHVRUKOFIRIK-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathryn A Dooley et al.
The Analyst, 138(17), 4838-4848 (2013-06-27)
In situ chemical imaging techniques are being developed to provide information on the spatial distribution of artists' pigments used in polychrome works of art such as paintings. The new methods include reflectance imaging spectroscopy and X-ray fluorescence mapping. Results from
Massimo Peruffo et al.
Physical chemistry chemical physics : PCCP, 15(6), 1956-1965 (2012-12-22)
Dissolution processes at single crystal surfaces often involve the initial formation and expansion of localized, characteristic (faceted) etch-pits at defects, in an otherwise comparatively unreactive surface. Using natural gypsum single crystal as an example, a simple but powerful morphological analysis
Joon Im et al.
American journal of orthodontics and dentofacial orthopedics : official publication of the American Association of Orthodontists, its constituent societies, and the American Board of Orthodontics, 145(4), 434-442 (2014-04-08)
The aim of this study was to compare the virtual and manual tooth setups with digital and plaster models in extraction cases by measuring various occlusal parameters and applying the American Board of Orthodontics objective grading system. Linear intra-arch and
Eun Young Rha et al.
Plastic and reconstructive surgery, 133(1), 14-20 (2014-01-01)
The authors have developed a simple and intuitive method for measuring breast volume based on three-dimensional simulated images of magnetic resonance imaging scans to accurately estimate breast volume before breast reconstruction. The authors performed a retrospective review of the medical
Nathan Evaniew et al.
Orthopedics, 36(2), e216-e222 (2013-02-06)
Benign primary bone tumors are commonly treated with intralesional curettage with or without the use of surgical adjuvants. The reconstructive approach to the resulting contained bone defects is controversial, and clinical practice is varied. Synthetic bone substitutes may provide early

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service