Skip to Content
Merck
All Photos(1)

Key Documents

90570

Sigma-Aldrich

Triethyl(trifluoromethyl)silane

purum, ≥98.0% (GC)

Synonym(s):

(Trifluoromethyl)triethylsilane, Triethylsilyl trifluoromethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3SiCF3
CAS Number:
Molecular Weight:
184.27
Beilstein:
4242161
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

grade

purum

Assay

≥98.0% (GC)

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.382 (lit.)
n20/D 1.382

bp

56-57 °C/60 mmHg (lit.)

density

0.98 g/mL at 25 °C (lit.)

SMILES string

CC[Si](CC)(CC)C(F)(F)F

InChI

1S/C7H15F3Si/c1-4-11(5-2,6-3)7(8,9)10/h4-6H2,1-3H3

InChI key

ZHSKFONQCREGOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Trifluoromethylation of aryl iodides
  • Trialkylsilylation reactions

  • Used for deposition of perfluoro-methyl silica films

Other Notes

Reagent used for fluoride-induced nucleophilic trifluoromethylations. The higher boiling point of this comp. allows reactions at higher temperatures than with the tri-methyl analogue

replaced by

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. Urata et al.
Tetrahedron Letters, 32, 91-91 (1991)
G.P. Stahly et al.
The Journal of Organic Chemistry, 54, 2873-2873 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service