Skip to Content
Merck
All Photos(1)

Key Documents

89953

Supelco

Dimethomorph-(dimethoxy-d6)

PESTANAL®, analytical standard

Synonym(s):

3-(4-Chlorophenyl)-3-[3,4-di(methoxy-d3)phenyl]-1-(4-morpholinyl)-2-propen-1-one, Dimethomorph-d6

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$392.00

$392.00


Estimated to ship onApril 28, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$392.00

About This Item

Empirical Formula (Hill Notation):
C21D6H16ClNO4
Molecular Weight:
393.89
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$392.00


Estimated to ship onApril 28, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

storage temp.

2-8°C

Related Categories

General description

Dimethomorph-(dimethoxy-d6) is an isotope-labeled analog of the systemic morpholine fungicide dimethomorph, wherein the dimethoxy protons are replaced by deuterium.[1]

Application

Dimethomorph-(dimethoxy-d6) may be used as a deuterated internal standard to quantify the analyte in dairy milk by liquid chromatography-tandem mass spectrometry (LC-MS/MS) technique.[2]
Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Hazard Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Responses of indigenous microorganisms to a fungicidal mixture of mancozeb and dimethomorph added to sandy soils
Cycon M, et al.
International Biodeterioration & Biodegradation, 64(4), 316-323 (2010)
Human pharmaceutical and pesticide residues in Israeli dairy milk in association with dietary risk assessment
Bommuraj V, et al.
Food additives & contaminants. Part B, Surveillance, 64(4), 1-11 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service