Skip to Content
Merck
All Photos(1)

Documents

78929

Sigma-Aldrich

(S)-2-Phenyl-1-propanol

purum, ≥98.0% (sum of enantiomers, GC)

Synonym(s):

(S)-β-Methylphenethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(C6H5)CH2OH
CAS Number:
Molecular Weight:
136.19
Beilstein:
2500176
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Assay

≥98.0% (sum of enantiomers, GC)

form

liquid

optical activity

[α]20/D −15.0±1°, c = 1% in toluene

refractive index

n20/D 1.526 (lit.)
n20/D 1.527

bp

214 °C (lit.)

density

1 g/mL at 25 °C (lit.)

SMILES string

C[C@H](CO)c1ccccc1

InChI

1S/C9H12O/c1-8(7-10)9-5-3-2-4-6-9/h2-6,8,10H,7H2,1H3/t8-/m1/s1

InChI key

RNDNSYIPLPAXAZ-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Goenechea et al.
Zeitschrift fur Rechtsmedizin. Journal of legal medicine, 97(2), 83-88 (1986-01-01)
The behaviour of 2-phenyl-1-propanol (I) and 2-phenyl-2-propanol (II) and their glucuronides with HCl has been investigated. While I shows a high acidic constancy, II undergoes a partial conversion into 2-phenylpropane (III) which itself yields numerous products. The glucosidic bond of
W Gielsdorf et al.
Zeitschrift fur Rechtsmedizin. Journal of legal medicine, 89(3), 191-195 (1982-01-01)
The FAB (Fast Atom Bombardment)-mass spectrometric ionization technique, which has now been available for about 1 year, has been successfully employed in forensic toxicology. The mass spectral behaviour of some representative drug-glucuronides (Codeine, p-Nitrophenol and 2-Phenyl-1-propanol) were studied by positive-
I F Gaunt et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 20(5), 519-525 (1982-10-01)
Phenylpropan-1-ol was added to the diet of groups of 15 male and 15 female rats to provide intakes of 0 (control), 10, 40 or 160 mg/kg/day for 13 wk. No effects on body weight, food intake, water intake, haematology, semi-quantitative
K R Henne et al.
Biochemistry, 40(29), 8597-8605 (2001-07-18)
The active site topography of rabbit CYP4B1 has been studied relative to CYP2B1 and CYP102 using a variety of aromatic probe substrates. Oxidation of the prochiral substrate cumene by CYP4B1, but not CYP2B1 or CYP102, resulted in the formation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service