Skip to Content
Merck
All Photos(1)

Key Documents

73608

Millipore

BiGGY Agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Bismuth Glycine Glucose Yeast Agar, Candida Selective Agar acc. to Nickerson, Nickerson Agar

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$311.00

$311.00


Estimated to ship onApril 18, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$311.00

About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

$311.00


Estimated to ship onApril 18, 2025


Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 16.0 g/L
bismuth ammonium citrate, 5.0 g/L
dextrose, 10.0 g/L
glycine, 10.0 g/L
sodium sulfite, 3.0 g/L
yeast extract, 1.0 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

6.8±0.2 (25 °C)

application(s)

agriculture
cleaning products
clinical testing
cosmetics
environmental
food and beverages
personal care
veterinary

microbiology

suitability

selective and differential for Candida spp.

General description

BiGGY Agar (Nickerson Agar) is used for detection, selective isolation, differentiation and presumptive identification of Candida albicans and Candida tropicalis. This agar is a selective medium used for isolation and presumptive identification of Candida species by their ability to reduce sulphite to sulphide giving a brown to black colour.

Application

BiGGY Agar has been used to evaluate the production of hydrogen sulfide in Saccharomyces cerevisiae.[1][2]

Preparation Note

Suspend 45 g in 1 litre distilled water. Boil gently to dissolve the medium completely. DO NOT AUTOCLAVE. Overheating will destroy the selective properties. Disperse the flocculant precipitate formed by swirling prior to dispensing into plates.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmaceutical Technology null
Ricardo Franco-Duarte et al.
Yeast (Chichester, England), 31(7), 265-277 (2014-04-23)
Genome sequencing is essential to understand individual variation and to study the mechanisms that explain relations between genotype and phenotype. The accumulated knowledge from large-scale genome sequencing projects of Saccharomyces cerevisiae isolates is being used to study the mechanisms that
Inês Mendes et al.
PloS one, 8(7), e66523-e66523 (2013-07-23)
Saccharomyces cerevisiae strains from diverse natural habitats harbour a vast amount of phenotypic diversity, driven by interactions between yeast and the respective environment. In grape juice fermentations, strains are exposed to a wide array of biotic and abiotic stressors, which

Articles

Yeast is one of the most important microorganisms known and utilised by mankind. Ancient Middle Eastern civilisations used the organism to bake bread and to produce mead, beer and wine.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service