Skip to Content
Merck
All Photos(1)

Documents

70127

Sigma-Aldrich

Naloxone hydrochloride dihydrate

≥99.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21NO4 · HCl · 2H2O
CAS Number:
Molecular Weight:
399.87
Beilstein:
6260807
EC Number:
MDL number:
UNSPSC Code:
12352202

Assay

≥99.0% (TLC)

storage temp.

2-8°C

SMILES string

O.O.Cl.Oc1ccc2C[C@H]3N(CC[C@@]45[C@@H](Oc1c24)C(=O)CC[C@@]35O)CC=C

InChI

1S/C19H21NO4.ClH.2H2O/c1-2-8-20-9-7-18-15-11-3-4-12(21)16(15)24-17(18)13(22)5-6-19(18,23)14(20)10-11;;;/h2-4,14,17,21,23H,1,5-10H2;1H;2*1H2/t14-,17+,18+,19-;;;/m1.../s1

InChI key

TXMZWEASFRBVKY-IOQDSZRYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Displaces morphine and other opiate agonists from the opiate receptor. In studies on the opiate receptor

Other Notes

Potent opiate antagonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Costa et al.
Proceedings of the National Academy of Sciences of the United States of America, 86(19), 7321-7325 (1989-10-01)
According to classical models of drug-receptor interactions, competitive antagonists share with agonists the ability to bind to a common site on the receptor molecule. However, they are different from agonists, as they cannot trigger the "stimulus" that leads to biological
C Mollereau et al.
The Journal of biological chemistry, 263(34), 18003-18008 (1988-12-05)
When assayed for specific opiate binding in the presence of 120 mM NaCl, digitonin extracts from frog (Rana ridibunda) brain membranes were found to contain about the same quantity (0.5 pmol/mg of protein) of high (Kdh = 0.4 nM) and
Yumei Liu et al.
PloS one, 8(1), e51573-e51573 (2013-02-06)
Previously, we demonstrated that electroacupuncture (EA) decreased lymphocyte infiltration into the spinal cords of rats presenting with experimental autoimmune encephalomyelitis (EAE), a disease model used in the study of multiple sclerosis (MS). The aim of this study was to characterize
Naloxone in the refractory neonatal septic shock.
M Radicioni et al.
Minerva pediatrica, 65(1), 122-123 (2013-02-21)
Opiate receptor antagonists for treatment of severe pruritus associated with advanced cholestatic liver disease.
Nilay Kumar et al.
Journal of palliative medicine, 16(2), 122-123 (2013-02-09)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service