Skip to Content
Merck
All Photos(1)

Key Documents

70123

Millipore

Nutrient Broth

No. 2, NutriSelect® Basic, suitable for microbiology

Synonym(s):

Standard - Nutrient Broth

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41171607

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

casein peptone, 4.3 g/L
meat peptone, 4.3 g/L
sodium chloride, 6.4 g/L

manufacturer/tradename

NutriSelect® Basic

final pH

7.5±0.2 (25 °C)

application(s)

agriculture
environmental
food and beverages

microbiology

suitability

nonselective for Escherichia coli
nonselective for Listeria spp.
nonselective for Salmonella spp.
nonselective for Shigella spp.
nonselective for Staphylococcus spp.
nonselective for Streptococcus spp.
nonselective for bacteria (General Media)
nonselective for coliforms

Looking for similar products? Visit Product Comparison Guide

Application

For the cultivation and enrichment of less fastidious bacteria for the establishment of permanent cultures, as well as a base in the preparation of special media.

Legal Information

NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service