Skip to Content
Merck
All Photos(1)

Key Documents

46021

Sigma-Aldrich

4-Nitrophenyl acetate

≥99.0% (GC)

Synonym(s):

Acetic acid 4-nitrophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C6H4NO2
CAS Number:
Molecular Weight:
181.15
Beilstein:
515874
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

≥99.0% (GC)

form

powder or crystals

mp

75-77 °C (lit.)
77-79 °C

storage temp.

−20°C

SMILES string

CC(=O)Oc1ccc(cc1)[N+]([O-])=O

InChI

1S/C8H7NO4/c1-6(10)13-8-4-2-7(3-5-8)9(11)12/h2-5H,1H3

InChI key

QAUUDNIGJSLPSX-UHFFFAOYSA-N

Gene Information

human ... PON1(5444)

Looking for similar products? Visit Product Comparison Guide

Other Notes

Activated acetate; reagent for acetylations. Substrate for carboxy-esterase[1]; Substrate for the determination of lipase[2]

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 3 - Skin Sens. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C S Wang
The Journal of biological chemistry, 256(19), 10198-10202 (1981-10-10)
Further studies on human milk bile salt-activated lipase were performed to provide kinetic and additional chemical characterizations of this enzyme. The enzyme was homogeneous by urea-sodium dodecyl sulfate-polyacrylamide gel electrophoresis and isoelectric focusing with an isoelectric point of 3.7. A
Multiple forms of carboxylesterases in Baker's yeast.
E Parkkinen
Cellular and molecular biology, including cyto-enzymology, 26(2), 147-154 (1980-01-01)
Conformer selection and intensified dynamics during catalytic turnover in chymotrypsin.
Peter Liuni et al.
Angewandte Chemie (International ed. in English), 51(38), 9666-9669 (2012-09-01)
Hanna Gustafsson et al.
Colloids and surfaces. B, Biointerfaces, 100, 22-30 (2012-07-04)
Immobilization of enzymes usually improves the recyclability and stability and can sometimes also improve the activity compared to enzymes free in solution. Mesoporous silica is a widely studied material as host for immobilized enzymes because of its large internal surface
Akın Akıncıoğlu et al.
Bioorganic & medicinal chemistry, 21(6), 1379-1385 (2013-02-12)
Sulfamides represent an important class of biologically active compounds. A series of novel sulfamides were synthesized from 1-aminoindanes, 1-aminotetralin, 2-aminoindanes and 2-aminotetralin via the reactions of free amines, benzyl alcohol and chlorosulfonyl isocyanate (CSI) followed by hydrogenolysis of the obtained

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service