Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44577

Sigma-Aldrich

Doxycycline hyclate

≥98.0% (TLC)

Synonym(s):

Doxycycline hydrochloride hemiethanolate hemihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O
CAS Number:
Molecular Weight:
512.94
Beilstein:
5702728
MDL number:
UNSPSC Code:
51101500
PubChem Substance ID:

biological source

synthetic

Assay

≥98.0% (TLC)

form

powder

impurities

≤5% water
≤7% ethanol

color

faintly yellow to yellow

solubility

H2O: 50 mg/mL, clear, yellow-green

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycoplasma
parasites

Mode of action

protein synthesis | interferes

SMILES string

Cl[H].Cl[H].[H]O[H].CCO.C[C@@H]1C2[C@H](O)C3[C@H](N(C)C)C(O)=C(C(N)=O)C(=O)[C@@]3(O)C(O)=C2C(=O)c4c(O)cccc14.C[C@@H]5C6[C@H](O)C7[C@H](N(C)C)C(O)=C(C(N)=O)C(=O)[C@@]7(O)C(O)=C6C(=O)c8c(O)cccc58

InChI

1S/2C22H24N2O8.C2H6O.2ClH.H2O/c2*1-7-8-5-4-6-9(25)11(8)16(26)12-10(7)17(27)14-15(24(2)3)18(28)13(21(23)31)20(30)22(14,32)19(12)29;1-2-3;;;/h2*4-7,10,14-15,17,25,27-29,32H,1-3H3,(H2,23,31);3H,2H2,1H3;2*1H;1H2/t2*7-,10?,14?,15-,17-,22-;;;;/m00..../s1

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: tetracycline

Biochem/physiol Actions

Mode of Action: Doxycycline hyclate inhibits the inflammatory response to the Lyme Disease Spirochete Borrelia burgdorferi.

Activity Spectrum: Effective against a broad spectrum inhibitor of matrix metalloproteinases in vivo.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.
This product has been replaced by D9891-SIGMA | Doxycycline hyclate ≥98% (TLC)

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Customers Also Viewed

    1. Which document(s) contains shelf-life or expiration date information for a given product?

      If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.

    2. How do I get lot-specific information or a Certificate of Analysis?

      The lot specific COA document can be found by entering the lot number above under the "Documents" section.

    3. How do I find price and availability?

      There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers.

    4. What is the Department of Transportation shipping information for this product?

      Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product. 

    5. Is doxycycline hyclate soluble in DMSO?

      According to literature sources, doxycycline hyclate is soluble in DMSO at a concentration of 100 mM.

    6. My question is not addressed here, how can I contact Technical Service for assistance?

      Ask a Scientist here.

    Anuradha Rajendran et al.
    The Journal of physiology, 598(22), 5109-5132 (2020-08-26)
    LAT4 is a broadly expressed uniporter selective for essential branched chain amino acids, methionine and phenylalanine, which are involved in epithelial transport. Its global deletion leads to an early malnutrition-like phenotype and death within 10 days after birth. Here, we
    Xuefeng Wu et al.
    Proceedings of the National Academy of Sciences of the United States of America, 111(38), 13870-13875 (2014-09-06)
    Metastatic spread is the leading cause of cancer mortality. Breast cancer (BCa) metastatic recurrence can happen years after removal of the primary tumor. Here we show that Ubc13, an E2 enzyme that catalyzes K63-linked protein polyubiquitination, is largely dispensable for
    Alicia Anderson et al.
    MMWR. Recommendations and reports : Morbidity and mortality weekly report. Recommendations and reports, 62(RR-03), 1-30 (2013-03-29)
    Q fever, a zoonotic disease caused by the bacterium Coxiella burnetii, can cause acute or chronic illness in humans. Transmission occurs primarily through inhalation of aerosols from contaminated soil or animal waste. No licensed vaccine is available in the United
    Alistair C Darby et al.
    The ISME journal, 8(4), 925-937 (2013-10-25)
    The bacterium Wolbachia (order Rickettsiales), representing perhaps the most abundant vertically transmitted microbe worldwide, infects arthropods and filarial nematodes. In arthropods, Wolbachia can induce reproductive alterations and interfere with the transmission of several arthropod-borne pathogens. In addition, Wolbachia is an
    Hanumara Ram Chowdri et al.
    Annals of internal medicine, 159(1), 21-27 (2013-07-03)
    The diverse tickborne infections of the northeastern United States can present as undifferentiated flu-like illnesses. In areas endemic for Lyme and other tickborne diseases, patients presenting with acute febrile illness with myalgia, headache, neutropenia, thrombocytopenia, and elevated hepatic aminotransferase levels

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service