Skip to Content
Merck
All Photos(1)

Key Documents

201308

Sigma-Aldrich

Pyronin B

certified by the Biological Stain Commission, Dye content 40 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H54Cl8Fe2N4O2
CAS Number:
Molecular Weight:
1042.22
Colour Index Number:
45010
Beilstein:
3846969
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

grade

certified by the Biological Stain Commission

form

solid

composition

Dye content, 40%

mp

174-176 °C (lit.)

λmax

553 nm

SMILES string

[Cl-].Cl[Fe](Cl)Cl.CCN(CC)c1ccc2cc3ccc(cc3[o+]c2c1)N(CC)CC

InChI

1S/C21H27N2O.4ClH.Fe/c1-5-22(6-2)18-11-9-16-13-17-10-12-19(23(7-3)8-4)15-21(17)24-20(16)14-18;;;;;/h9-15H,5-8H2,1-4H3;4*1H;/q+1;;;;;+3/p-4

InChI key

SUVPQRYDMCNTPV-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Suitability

Certified for use in the methyl green-pyronin method for differential coloration of nucleic acids.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nuclear dyes and cytoplasmic staining.
G Diaz et al.
Cytometry, 29(4), 375-376 (1998-01-24)
Feng Gao et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 60(11), 2505-2509 (2004-08-06)
This is the first report on the determination of nucleic acids with Pyronine B (PB) sensitized by cetyltrimethylammonium bromide (CTMAB) with resonance light-scattering (RLS) technique. Under the experimental conditions (1 x 10(-5) mol l(-1) PB, 1 x 10(-5) mol l(-1)
Behzad Rezaei et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 66(4-5), 869-873 (2006-07-29)
A new simple, selective, high sensitive and rapid method has been developed for spectrophotometric determination of ultra trace amounts of ruthenium based on its catalytic effect on the oxidation of pyronin B by periodate at lambdamax=555 nm. The described method
T Frey
Cytometry, 21(3), 265-274 (1995-11-01)
Apoptotic thymocytes were found to be much dimmer than normal thymocytes when stained with several nucleic acid dyes. These dyes provide a quick and simple assay for apoptosis which works for live cells and does not require a UV laser.
E K Paleologos et al.
Analytical chemistry, 73(18), 4428-4433 (2001-09-29)
The catalytic effect of vanadium on the pyronine B-H2O2 system is examined. Enhancement of the catalytic reaction rate along with the efficiency and selectivity against vanadium is achieved in a formic acid environment in the presence of a nonionic surfactant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service