Skip to Content
Merck
All Photos(1)

Documents

28-3100

Sigma-Aldrich

Sodium hypochlorite solution

CP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaClO
CAS Number:
Molecular Weight:
74.44
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

CP

form

liquid

composition

available chlorine, ~10%

reaction suitability

reagent type: oxidant

availability

available only in Japan

storage temp.

15-25°C

SMILES string

[Na+].[O-]Cl

InChI

1S/ClO.Na/c1-2;/q-1;+1

InChI key

SUKJFIGYRHOWBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Poly(pyridinium salt)s with organic counterions derived from 3,3′-dimethylnaphthidine: thermal, liquid crystalline, and optical properties.: This research explores the thermal and optical properties of poly(pyridinium salt)s derived from 3,3′-dimethylnaphthidine, highlighting their potential in creating materials with unique electro-optical properties suitable for advanced applications in displays and photovoltaics (Jose et al., 2015).



Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anthony A Chariton et al.
Environmental pollution (Barking, Essex : 1987), 203, 165-174 (2015-04-25)
DNA-derived measurements of biological composition have the potential to produce data covering all of life, and provide a tantalizing proposition for researchers and managers. We used metabarcoding to compare benthic eukaryote composition from five estuaries of varying condition. In contrast
Amy C Degnim et al.
Annals of surgery, 258(2), 240-247 (2013-03-23)
To determine whether bacterial colonization of drains can be reduced by local antiseptic interventions. Drains are a potential source of bacterial entry into surgical wounds and may contribute to surgical site infection after breast surgery. After institutional review board approval
Bin Chang et al.
Applied and environmental microbiology, 75(1), 147-153 (2008-11-04)
Legionella organisms are prevalent in manmade water systems and cause legionellosis in humans. A rapid detection method for viable Legionella cells combining ethidium monoazide (EMA) and PCR/real-time PCR was assessed. EMA could specifically intercalate and cleave the genomic DNA of
Kyle L Brown et al.
Diabetes, 64(6), 2242-2253 (2015-01-22)
In diabetes, toxic oxidative pathways are triggered by persistent hyperglycemia and contribute to diabetes complications. A major proposed pathogenic mechanism is the accumulation of protein modifications that are called advanced glycation end products. However, other nonenzymatic post-translational modifications may also
Sarah L Edmonds et al.
Infection control and hospital epidemiology, 34(3), 302-305 (2013-02-08)
This study determined whether surrogate organisms can predict activity against Clostridium difficile spores and compared the efficacy of hand hygiene preparations against C. difficile. Our data suggest that surrogate organisms were not predictive of C. difficile spore removal. Four preparations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service