Skip to Content
Merck
All Photos(1)

Documents

09-1661

Sigma-Aldrich

Ethylene glycol butyl ether

SAJ first grade, ≥99.0%

Synonym(s):

2-Butoxyethanol, Butyl glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OCH2CH2OH
CAS Number:
Molecular Weight:
118.17
Beilstein:
1732511
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:

grade

SAJ first grade

vapor density

4.1 (vs air)

vapor pressure

<1 mmHg ( 20 °C)
10 mmHg ( 81 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

473 °F

expl. lim.

10.6 %

availability

available only in Japan

refractive index

n20/D 1.419 (lit.)

bp

169-172.5 °C (lit.)

mp

−75 °C (lit.)

density

0.902 g/mL at 25 °C (lit.)

SMILES string

CCCCOCCO

InChI

1S/C6H14O2/c1-2-3-5-8-6-4-7/h7H,2-6H2,1H3

InChI key

POAOYUHQDCAZBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

152.6 °F - Pensky-Martens closed cup

Flash Point(C)

67 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prajakta S Palkar et al.
Toxicology and applied pharmacology, 230(3), 338-345 (2008-04-26)
Administration of a low priming dose of 2-butoxyethanol (BE, 500 mg/kg, p.o.) 7 days prior to a larger LD(90) dose (1500 mg BE/kg, p.o.) offers protection against the lethal dose-induced hemolysis and death in female Sprague Dawley rats because of
R Rella et al.
Journal of environmental monitoring : JEM, 14(10), 2659-2662 (2012-08-18)
Indoor air quality is important because people are spending an increasing amount of time in the workplace. They are exposed to outdoor pollutants as well as pollutants emitted from products used indoors. Some chemicals, belonging to the category of volatile
G Amir et al.
The British journal of oral & maxillofacial surgery, 49(1), 21-25 (2009-12-26)
Female rats develop haemolytic anaemia and disseminated thrombosis and infarction in multiple organs, including bone, when exposed to 2-butoxyethanol (BE). There is growing evidence that vascular occlusion of the subchondral bone may play a part in some cases of osteoarthritis.
Ryszard Tokarczyk et al.
Journal of chromatography. A, 1217(44), 6964-6970 (2010-09-22)
A gas chromatography-mass spectrometry isotope dilution (GC-MS ID) method was developed and tested for the determination of 14 common glycol ethers in consumer products. Stable isotope labelled standards, 2-methoxyethanol-D(7) and 2-butoxyethanol-(13)C(2) (CDN isotopes) were employed to enhance the accuracy and
Matthew J Traynor et al.
Toxicology letters, 177(3), 151-155 (2008-03-04)
Glycol ethers are widely used in industrial and household applications because their chemical and physical properties make them versatile solvents, miscible with both water and organic media. Due to the ease with which the glycol ethers are absorbed through the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service