Skip to Content
Merck
All Photos(1)

Documents

07-5560

Sigma-Aldrich

1,5-Diphenylcarbazide

JIS special grade

Synonym(s):

1,5-Diphenylcarbohydrazide, sym.-Diphenylcarbazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHNHCONHNHC6H5
CAS Number:
Molecular Weight:
242.28
Beilstein:
752039
EC Number:
MDL number:
UNSPSC Code:
12352127
PubChem Substance ID:

grade

JIS special grade

availability

available only in Japan

mp

170-175 °C (lit.)

SMILES string

O=C(NNc1ccccc1)NNc2ccccc2

InChI

1S/C13H14N4O/c18-13(16-14-11-7-3-1-4-8-11)17-15-12-9-5-2-6-10-12/h1-10,14-15H,(H2,16,17,18)

InChI key

KSPIHGBHKVISFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B K Semin et al.
FEBS letters, 375(3), 223-226 (1995-11-20)
The interaction of Fe(II) and Fe(III) with the 'high-affinity Mn-binding site' in Mn-depleted photosystem II (PSII) was investigated using diphenilcarbazide (DPC)/2,6-dichlorophenol-indophenol (DCIP) inhibition assay. Fe(III) was ineffective in the inhibition of DPC-DCIP reaction while Fe(II) decreased the rate of DCIP
N Rajesh et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 69(2), 612-618 (2007-07-03)
A novel approach has been developed for the solid phase extraction of chromium(VI) based on the adsorption of its diphenylcarbazide complex on a mixture of acid activated montmorillonite (AAM)-silica gel column. The effect of various parameters such as acidity, stability
M L Ghirardi et al.
Biochemistry, 35(6), 1820-1828 (1996-02-13)
The inhibition of DPC-mediated DCIP photoreduction by exogenous MnCl2 in Tris-treated photosystem II (PSII) membrane fragments has been used to probe for amino acids on the PSII reaction center proteins, including D1His337, that provide ligands for binding manganese [Preston, C.
Akbar Bagheri et al.
Talanta, 89, 455-461 (2012-01-31)
A novel sorbent for simultaneous separation of cadmium and copper was prepared by functionalizing SBA-15 nanoporous silica with diphenylcarbazide. A solid-phase extraction method using the above sorbent has been developed to separate and concentrate trace amount of cadmium and copper
M L Ghirardi et al.
Biochemistry, 37(39), 13559-13566 (1998-09-30)
Our previous work using the "diphenylcarbazide (DPC)-inhibition assay" has identified four amino acid (two carboxyls and two histidyls) ligands to four Mn2+ bound with high affinity on Tris-washed photosystem II (PSII) membrane fragments [Preston and Seibert (1991) Biochemistry 30, 9615-9624

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service