01-5150
Ammonium sulfide solution
yellow, SAJ first grade, 5-6% S
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
SAJ first grade
form
liquid
availability
available only in Japan
concentration
5-6% S
color
yellow
SMILES string
N.N.S
InChI
1S/2H3N.H2S/h2*1H3;1H2
InChI key
UXXGWUQNRMPNKH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of agricultural and food chemistry, 47(1), 245-248 (1999-11-24)
The reactions between 3-hydroxy-2-butanone and ammoniun sulfide at 25, 50, 75, 100, 125, and 150 degrees C were studied. Four well-known flavor compounds, 2,4,5-trimethyloxazole, 2,4, 5-trimethyl-3-oxazoline, 2,4,5-trimethylthiazole, and 2,4, 5-trimethyl-3-thiazoline, were identified. Another four interesting intermediate compounds, 2-(1-hydroxyethyl)-2,4, 5-trimethyl-3-oxazoline, 2-(1-mercaptoethyl)-2,4
Chemical senses, 35(1), 31-40 (2009-11-18)
To assess the feasibility of using odors as a potential mechanism for treating sleep apnea, we set out to test the hypothesis that odorants delivered during sleep would modify respiratory patterns without inducing arousal or wake in healthy sleepers. We
The journal of physical chemistry. B, 110(29), 14398-14405 (2006-07-21)
This work studies the evolution of 1018 carbon steel surfaces during 3-15 day immersion in alkaline sour medium 0.1 M (NH4)2S and 10 ppm CN(-) as (NaCN). During this period of time, surfaces were jointly characterized by electrochemical techniques in
[Photographic models in the study and valuation of the silver reactions in histology. II. Optical density variation in enlarging exposure scale under the action of the ammonium sulphide on silver (author's transl)].
Mikroskopie, 36(3-4), 117-127 (1980-06-01)
Luminescence : the journal of biological and chemical luminescence, 25(4), 294-299 (2009-07-07)
In the present work, Fe(3)O(4)-carbon nanotubes (CNTs) composite was explored as a sensing material candidate for ammonium sulfide. Intense chemiluminescence emission can be observed during the catalytic oxidation of ammonium sulfide on the surface of Fe(3)O(4)-CNTs composite. Based on this
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service