Skip to Content
Merck

Skip To

DNTP-RO

Roche

Deoxynucleoside Triphosphate Set

PCR Grade, sodium salt

Synonym(s):

datp, dctp, dgtp, dttp, set of datp, dctp, dgtp, and dttp, pcr grade

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
4 x 250 μL

Estimated to ship onAugust 28, 2026fromCarlos Spegazzini

$250.00
4 x 1250 μL

Estimated to ship onAugust 28, 2026fromCarlos Spegazzini

$1,440.00

About This Item

NACRES:
NA.21
UNSPSC Code:
41106300
Assay:
99% (dNTP)
Form:
liquid
Solubility:
water: miscible
Storage temp.:
−20°C

$250.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onAugust 28, 2026Details


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

assay

99% (dNTP)

form

liquid

usage

sufficient for 37,500 reactions (03622614001), sufficient for 6,250 reactions (11969064001)

packaging

pkg of 4 × 1,250 μL (03622614001 [4x100 mM]), pkg of 4 × 250 μL (11969064001 [4x100 mM])

manufacturer/tradename

Roche

concentration

<0.9% (dNDP)

technique(s)

whole genome amplification: suitable

color

colorless

solubility

water: miscible

storage temp.

−20°C

General description

This product contains 4 separate vials of dATP, PCR Grade, dCTP, PCR Grade, dGTP, PCR Grade, and dTTP, PCR Grade, each with a concentration of 100 mM and as a clear, colorless solution of the sodium salt (pH 8.3).

Application

The Deoxynucleoside Triphosphate Set, PCR Grade, is suitable for many applications where high-quality reagents are required. Such applications include reverse transcription (RT), polymerase chain reaction (PCR), RT-PCR, DNA labeling reactions, and sequencing/cycle sequencing analysis.[1][2]

Features and Benefits

PCR Grade Nucleotides from Roche are specially manufactured and purified to the highest possible chemical purity.
  • PCR Grade Nucleotides contain at least 99% of the relevant deoxynucleoside-triphosphate (dNTP) and less than 0.9% deoxynucleoside-diphosphate (dNDP).
  • They are provided as clear, colorless solutions in water at a concentration of 100 mM (pH 8.3).
  • Due to the manufacturing process, PCR Grade Nucleotides are free of dNTPs with modified bases, as well as tetra- and pyrophosphate contamination. Tetraphosphates can disintegrate and form pyrophosphate, which is a known inhibitor of amplification reactions. Therefore, the purity of nucleotides is critical to the success of your amplification reactions.
  • PCR Grade Nucleotides are stable for 30 months after the production date when stored at -15 to -25°C.

Packaging

1 set containing 4 components
  • dATP, PCR Grade, sodium salt, 100 mM
  • dCTP, PCR Grade, sodium salt, 100 mM
  • dGTP, PCR Grade, sodium salt, 100 mM
  • dTTP, PCR Grade, sodium salt, 100 mM

Preparation Note

Working solution: For best results, prepare a deoxynucleotide mix containing dATP, dCTP, dGTP, dTTP (10 mM, each); (e.g., for the preparation of 100 μl nucleotide mix add 10 μl of dATP, dCTP, dGTP, dTTP (each) to 60 μl Water, PCR Grade).

Analysis Note

Function tested using RT-PCR: Tested for absence of contaminating deoxyribonucleases, ribonucleases, and nicking activities according to the current Quality Control procedures.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
03732738001DNSTP-RODNTP100A
description

PCR Grade, sodium salt

description

Sequencing Grade, sodium salt

description

≤1.5% (dNDP, HPLC), ≥=98% (dNTP), ≤0.5% (dNMP, HPLC)

description

Individual dNTPs for routine PCR; 1 mL each

grade

Molecular Biology

grade

Molecular Biology

grade

-

grade

-

manufacturer/tradename

Roche

manufacturer/tradename

Roche

manufacturer/tradename

Roche

manufacturer/tradename

-

form

liquid

form

liquid

form

liquid

form

liquid

assay

99% (dNTP)

assay

≤1.5% (ddNDP, HPLC), ≥98% (ddNTP, HPLC)

assay

≥98% (dntp)

assay

-

storage temp.

−20°C

storage temp.

−20°C (−15°C to −25°C)

storage temp.

−20°C

storage temp.

−20°C

solubility

water: miscible

solubility

water: miscible

solubility

water: miscible

solubility

-


Still not finding the right product?

Explore all of our products under Deoxynucleoside Triphosphate Set


Kit Components Only

Product No.
Description

  • dATP, PCR Grade, sodium salt 100 mM

  • dCTP, PCR Grade, sodium salt 100 mM

  • dGTP, PCR Grade, sodium salt 100 mM

  • dTTP, PCR Grade, sodium salt 100 mM

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library