Skip to Content
Merck
All Photos(1)

Key Documents

SX1244P

Supelco

Sulfuric Acid Solution for Babcock Milk Test

for Babcock Milk Test

Synonym(s):

Sulfuric acid solution, acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
41116116
EC Index Number:
231-639-5

vapor pressure

0.0001 hPa ( 20 °C)

Quality Level

Assay

90.5-92.8%

form

liquid

shelf life

24 mo. (from manufacture date)

technique(s)

Babcock Milk Test: suitable

color

APHA: ≤30

pH

0.3 (25 °C, 49 g/L in H2O)

bp

335 °C/1013 hPa

mp

-20 °C

shipped in

ambient

storage temp.

no temp limit

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

for Babcock Milk Test

Analysis Note

Assay90.5-92.8%Color (APHA)30 maxExpiration dateExp. 24 months from manufacturing dateFormClear liquidSpecific Gravity at 20/4°C1.820-1.830

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service