Skip to Content
Merck
All Photos(1)

Documents

SCM107

Sigma-Aldrich

Human OPC Expansion Culture Media Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32

description

Neural Stem Cells

Quality Level

type

for cell culture

form

liquid

technique(s)

cell culture | stem cell: suitable

General description

Human OPC Expansion Media is a defined, serum-free medium that has been optimized for the culture and expansion of human ES derived oligodendrocyte progenitor cells (Catalog No. SCR600). Cells double every 48 hours when cultured in Human OPC Expansion Complete Media. It is not known whether the media can support the culture of primary oligodendrocyte progenitor cells isolated from brain tissues.

Storage and Stability

Store at -20°C, good for 4 months from date of receipt when reagents are handled and stored appropriately. Please note: The supplemented Human OPC Expansion cOmplete Media should be stored at 2 to 8C for up to 4 weeks. .

Legal Information

cOmplete is a trademark of Roche

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service